Merck
  • Retrievability of implant-supported crowns when using three different cements: a controlled clinical trial.
Retrievability of implant-supported crowns when using three different cements: a controlled clinical trial.
The International journal of prosthodontics (2015-01-15)
Andreas Worni, Hadi Gholami, Laurent Marchand, Joannis Katsoulis, Regina Mericske-Stern, Norbert Enkling
ABSTRACT
The purpose of this study was to analyze the removal of implant-supported crowns retained by three different cements using an air-accelerated crown remover and to evaluate the patients' response to the procedure. This controlled clinical trial was conducted with 21 patients (10 women, 11 men; mean age: 51 ± 10.2 years) who had received a total of 74 implants (all placed in the posterior zone of the mandible). Four months after implant surgery, the crowns were cemented on standard titanium abutments of different heights. Three different cements (two temporary: Harvard TEMP and Improv; and one definitive: Durelon) were used and randomly assigned to the patients. Eight months later, one blinded investigator removed all crowns. The number of activations of the instrument (CORONAflex, KaVo) required for crown removal was recorded. The patients completed a questionnaire retrospectively to determine the impact of the procedure and to gauge their subjective perception. A linear regression model and descriptive statistics were used for data analysis. All crowns could be retrieved without any technical complications or damage. Both abutment height (P = .019) and cement type (P = .004) had a significant effect on the number of activations, but the type of cement was more important. An increased total number of activations had no or only a weak correlation to the patients' perception of concussion, noise, pain, and unwillingness to use the device. Cemented implant crowns can be removed, and the application of an air-accelerated device is a practicable method. A type of cement with appropriate retention force has to be selected. The impact on the patients' subjective perception should be taken into account.
Sigma-Aldrich
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Titanium, IRMM®, certified reference material, 0.5 mm foil
Titanium, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, ≤10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Titanium, tube, 1000mm, outside diameter 6.35mm, inside diameter 5.53mm, wall thickness 0.41mm, annealed, 99.6+%
Titanium, rod, 1000mm, diameter 1.5mm, annealed, 99.6+%
Titanium, mesh, 100x100mm, nominal aperture 0.19mm, wire diameter 0.23mm, 60x60 wires/inch, open area 20%, twill weave
Titanium, tube, 1000mm, outside diameter 1.6mm, inside diameter 1.2mm, wall thickness 0.2mm, hard, 99.6+%
Titanium, rod, 200mm, diameter 2mm, annealed, 99.6+%
Titanium, wire reel, 1m, diameter 0.20mm, annealed, 99.6+%
Titanium, tube, 500mm, outside diameter 6.35mm, inside diameter 4.57mm, wall thickness 0.89mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 6.1mm, inside diameter 5.1mm, wall thickness 0.5mm, annealed, 99.6+%
Titanium, tube, 1000mm, outside diameter 12.7mm, inside diameter 10.9mm, wall thickness 0.9mm, annealed, 99.6+%
Titanium, rod, 100mm, diameter 2mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 4.25mm, inside diameter 3.75mm, wall thickness 0.25mm, annealed, 99.6+%
Titanium, rod, 500mm, diameter 5mm, annealed, 99.6+%
Titanium, wire reel, 5m, diameter 0.5mm, as drawn, 99.6+%
Titanium, wire reel, 10m, diameter 0.125mm, annealed, 99.6+%
Titanium, tube, 500mm, outside diameter 2.03mm, inside diameter 1.55mm, wall thickness 0.24mm, annealed, 99.6+%
Titanium, tube, 500mm, outside diameter 0.81mm, inside diameter 0.61mm, wall thickness 0.1mm, as drawn, 99.6+%
Titanium, wire reel, 50m, diameter 0.75mm, as drawn, 99.6+%
Titanium, wire reel, 200m, diameter 0.125mm, annealed, 99.6+%
Titanium, wire reel, 25m, diameter 0.25mm, as drawn, 99.6+%
Titanium, tube, 500mm, outside diameter 25.4mm, inside diameter 23.62mm, wall thickness 0.89mm, annealed, 99.6+%
Titanium, wire reel, 1m, diameter 0.25mm, as drawn, 99.6+%