Merck
  • Artemin augments survival and axon regeneration in axotomized retinal ganglion cells.
Artemin augments survival and axon regeneration in axotomized retinal ganglion cells.
Journal of neuroscience research (2014-07-22)
Kazuko Omodaka, Takuji Kurimoto, Orie Nakamura, Kota Sato, Masayuki Yasuda, Yuji Tanaka, Noriko Himori, Yu Yokoyama, Toru Nakazawa
ABSTRACT
Artemin, a recently discovered member of the glial cell line-derived neurotrophic factor (GDNF) family, has neurotrophic effects on damaged neurons, including sympathetic neurons, dopamine neurons, and spiral ganglion neurons both in vivo and in vitro. However, its effects on retinal cells and its intracellular signaling remain relatively unexplored. During development, expression of GFRα3, a specific receptor for artemin, is strong in the immature retina and gradually decreases during maturation, suggesting a possible role in the formation of retinal connections. Optic nerve damage in mature rats causes levels of GFRα3 mRNA to increase tenfold in the retina within 3 days. GFRα3 mRNA levels continue to rise within the first week and then decline. Artemin, a specific ligand for GFRα3, has a neuroprotective effect on axotomized retinal ganglion cells (RGCs) in vivo and in vitro via activation of the extracellular signal-related kinase- and phosphoinositide 3-kinase-Akt signaling pathways. Artemin also has a substantial effect on axon regeneration in RGCs both in vivo and in vitro, whereas other GDNF family members do not. Therefore, artemin/GFRα3, but not other GDNF family members, may be of value for optic nerve regeneration in mature mammals.
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Anti-GFR α3 Antibody, serum, Chemicon®
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Trypan Blue solution, 0.4%, suitable for microscopy