Merck
  • An RNA-sequencing transcriptome and splicing database of glia, neurons, and vascular cells of the cerebral cortex.
An RNA-sequencing transcriptome and splicing database of glia, neurons, and vascular cells of the cerebral cortex.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2014-09-05)
Ye Zhang, Kenian Chen, Steven A Sloan, Mariko L Bennett, Anja R Scholze, Sean O'Keeffe, Hemali P Phatnani, Paolo Guarnieri, Christine Caneda, Nadine Ruderisch, Shuyun Deng, Shane A Liddelow, Chaolin Zhang, Richard Daneman, Tom Maniatis, Ben A Barres, Jian Qian Wu
ABSTRACT
The major cell classes of the brain differ in their developmental processes, metabolism, signaling, and function. To better understand the functions and interactions of the cell types that comprise these classes, we acutely purified representative populations of neurons, astrocytes, oligodendrocyte precursor cells, newly formed oligodendrocytes, myelinating oligodendrocytes, microglia, endothelial cells, and pericytes from mouse cerebral cortex. We generated a transcriptome database for these eight cell types by RNA sequencing and used a sensitive algorithm to detect alternative splicing events in each cell type. Bioinformatic analyses identified thousands of new cell type-enriched genes and splicing isoforms that will provide novel markers for cell identification, tools for genetic manipulation, and insights into the biology of the brain. For example, our data provide clues as to how neurons and astrocytes differ in their ability to dynamically regulate glycolytic flux and lactate generation attributable to unique splicing of PKM2, the gene encoding the glycolytic enzyme pyruvate kinase. This dataset will provide a powerful new resource for understanding the development and function of the brain. To ensure the widespread distribution of these datasets, we have created a user-friendly website (http://web.stanford.edu/group/barres_lab/brain_rnaseq.html) that provides a platform for analyzing and comparing transciption and alternative splicing profiles for various cell classes in the brain.
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
L-Cysteine hydrochloride monohydrate, reagent grade, ≥98% (TLC)
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Bovine Serum Albumin, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Anti-Neural Cell Adhesion Molecule L1 Antibody, clone 324, clone 324, Chemicon®, from rat
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
Fluorescein, suitable for fluorescence, free acid
SAFC
L-Cysteine
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Earle′s Balanced Salt Solution 10x, Without sodium bicarbonate, 10 ×, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
L-Cysteine, Wacker Chemie AG, ≥98.0%
Supelco
L-Cysteine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Fluorescein, European Pharmacopoeia (EP) Reference Standard