Merck
  • Discordant expression and variable numbers of neighboring GGA- and GAA-rich triplet repeats in the 3' untranslated regions of two groups of messenger RNAs encoded by the rat polymeric immunoglobulin receptor gene.
Discordant expression and variable numbers of neighboring GGA- and GAA-rich triplet repeats in the 3' untranslated regions of two groups of messenger RNAs encoded by the rat polymeric immunoglobulin receptor gene.
Nucleic acids research (1995-04-11)
K S Koch, A S Gleiberman, T Aoki, H L Leffert, A Feren, A L Jones, E J Fodor
ABSTRACT
An unusual S1-nuclease sensitive microsatellite (STMS) has been found in the single copy, rat polymeric immunoglobulin receptor gene (PIGR) terminal exon. In Fisher rats, elements within or beyond the STMS are expressed variably in the 3' untranslated regions (3'UTRs) of two 'Groups' of PIGR-encoded hepatic mRNAs (pIg-R) during liver regeneration. STMS elements include neighboring constant regions (a 60-bp d[GA]-rich tract with a chi-like octamer, followed by 15 tandem d[GGA] repeats) that merge directly with 36 or 39 tandem d[GAA] repeats (Fisher or Wistar strains, respectively) interrupted by d[AA] between their 5th-6th repeat units. The Wistar STMS is flanked upstream by two regions of nearly contiguous d[CA] or d[CT] repeats in the 3' end of intron 8; and downstream, by a 283 bp 'unit' containing several inversions at its 5' end, and two polyadenylation signals at its 3' end. The 283 nt unit is expressed in Group 1 pIg-R mRNAs; but it is absent in the Group 2 family so that their GAA repeats merge with their poly A tails. In contrast to genomic sequence, GGA triplet repeats are amplified (n > or = 24-26), whereas GAA triplet repeats are truncated variably (n < or = 9-37) and expressed uninterruptedly in both mRNA Groups. These results suggest that 3' end processing of the rat PIGR gene may involve misalignment, slippage and premature termination of RNA polymerase II. The function of this unusual processing and possible roles of chi-like octamers in quiescent or extrahepatic tissues are discussed.