Merck
  • Xanthine derivatives quantification in serum by capillary zone electrophoresis.
Xanthine derivatives quantification in serum by capillary zone electrophoresis.
Journal of chromatographic science (2013-11-14)
Juan Peris-Vicente, Maria Rambla-Alegre, Abhilasha Durgavanshi, Devasish Bose, Josep Esteve-Romero, Sergio Marco-Peiró
ABSTRACT
A capillary electrophoresis method was developed to quantify caffeine and theophylline, xanthine derivatives with bronchodilator activity. Buffer concentration, pH and applied voltage were optimized using a central composite design-face centred. Separation conditions were: silica capillary tube, 75 μm (i.d.) and 61 cm (total length); absorbance detection, 280 nm; borate buffer, 20 mM, pH 9.0; applied voltage, 25 kV and 1 psi injection/8 s. Validation was performed in blank serum following the International Conference Harmonization guidelines: resolution (peaks without overlapping), linear range (0.125-50 µg/mL; r(2) > 0.9999), limits of detection and quantification (10; 20 and 33; 66 ppb for caffeine and theophylline, respectively), intra- and inter-day precision (Relative standard deviation lower than 1.9%) and accuracy (98-101%). Migration times were <8 min. This method is simple, specific and suitable and reaches high label claims (98.7-100.4%) in pharmaceutical formulations analysis. Moreover, the method was applied to the monitoring of the analytes in serum of patients.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Boric acid-11B, ≥99 atom % 11B
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Supelco
Theophylline, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium acetate solution, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Theophylline, anhydrous, ≥99%, powder
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Supelco
Melting point standard 235-237°C, analytical standard
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O