Merck
  • Shengxian decoction in chronic heart failure treatment and synergistic property of Platycodonis Radix: a metabolomic approach and its application.
Shengxian decoction in chronic heart failure treatment and synergistic property of Platycodonis Radix: a metabolomic approach and its application.
Molecular bioSystems (2014-05-20)
Feng Zhang, Qin Zhan, Xin Dong, Bo Jiang, Lianna Sun, Shouhong Gao, Zhiqing He, Xia Tao, WanSheng Chen
ABSTRACT
Shengxian Decoction (SXT), a classic Traditional Chinese Medicine (TCM) prescription, consists of five TCMs: Astragali Radix, Anemarrhenae Rhizoma, Bupleuri Radix, Platycodonis Radix (PG), and Cimicifugae Rhizoma. SXT has been demonstrated to show good therapeutic effects on the cardiovascular system. A metabolomic approach was applied to study its therapeutic mechanisms and the synergistic properties of PG. UPLC-Q-TOF/MS based metabolomic profiling was adopted to assess the intervention effects of SXT, SXT-PG (SXT lacking PG) and PG, on chronic heart failure (CHF) rats. Betaloc was used as a positive control drug. A supervised discriminant technique (PLS-DA) was used to visualize the difference in global metabolic profiles within all experimental groups. Some significantly changed metabolites, such as carnitines, long-chain fatty acids and sphinganines, were identified, and the biochemical alterations of these were related to the disturbance in serum metabolic profiling of CHF rats. Furthermore, the metabolomics study demonstrated that the administration of SXT, but neither SXT-PG or PG alone, gave satisfactory curative effects on CHF through partially regulating the perturbed metabolic pathways. These observations were demonstrated by histopathological, electrocardiogram and serum enzymatic investigations. All these results supported the TCM theory that PG possesses synergistic properties which promote the synergized herbs in SXT in CHF rats. Overall, this paper demonstrates that metabolomics offers opportunities to understand the therapeutic mechanisms and synergistic properties of TCM.
Product Description
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammonium acetate solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, BioUltra, ≥99.0%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG