Merck
  • Cytoplasmic and nuclear toxicity of 3,5-dimethylaminophenol and potential protection by selenocompounds.
Cytoplasmic and nuclear toxicity of 3,5-dimethylaminophenol and potential protection by selenocompounds.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2014-07-12)
Pinar Erkekoglu, Ming-Wei Chao, Wenjie Ye, Jing Ge, Laura J Trudel, Paul L Skipper, Belma Kocer-Gumusel, Bevin P Engelward, Gerald N Wogan, Steven R Tannenbaum
ABSTRACT
Most common alkylanilines in the environment are 2,6-dimethylaniline (2,6-DMA), 3,5-dimethylaniline (3,5-DMA), and 3-ethylaniline (3-EA). 3,5-Dimethylaminophenol (3,5-DMAP), a metabolite of 3,5-DMA, is of particular interest, as it is potentially genotoxic. Supplementation with organic or inorganic forms of selenium (Se) may reduce toxicity following exposure to a wide variety of environmental chemicals. This study was designed to evaluate the protective effects of sodium selenite (SS) and selenomethionine (SM) at varying time points of supplementation (24 h and 72 h) against the cytotoxicity, reactive oxygen species (ROS) production, and genotoxicity of 3,5-DMAP in CHO AS52 cells. 3,5-DMAP caused dose-dependent increase of cytotoxicity, ROS production and genotoxicity, and generated free radicals in the nuclei. Thioredoxin reductase (TrxR), catalase and glutathione reductase activities, and glutathione levels were significantly lower while lipid peroxidation and protein oxidation levels were higher after 3,5-DMAP treatment in both cytoplasm and the nucleus vs. control. After 24 h, both SS and SM provided protection in antioxidant/oxidant status of the 3,5-DMAP-treated cells; however other than supplying higher glutathione peroxidase and TrxR activities, 72 h supplementation did not provide advanced improvement. Selenocompounds may be beneficial against cytotoxic and genotoxic potential of 3,5-DMAP and might protect both nucleus and cytoplasm following exposure to alkylanilines.
Product Description
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard
Trolamine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Ethyl methanesulfonate, liquid
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
6-Thioguanine, ≥98%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
6-Thioguanine, Hybri-Max, 50 ×, γ-irradiated, lyophilized powder, BioXtra, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Triethanolamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethanolamine, ≥99.0% (GC)
Sigma-Aldrich
Triethanolamine, puriss. p.a., ≥99% (GC)
Sigma-Aldrich
Triethanolamine, reagent grade, 98%
Sigma-Aldrich
2-Vinylpyridine, 97%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Triethanolamine, puriss., meets analytical specification of NF, ≥99% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Supelco
Ethyl methanesulfonate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Triethanolamine, analytical standard