Merck
  • Cu-TiO2 nanorods with enhanced ultraviolet- and visible-light photoactivity for bisphenol A degradation.
Cu-TiO2 nanorods with enhanced ultraviolet- and visible-light photoactivity for bisphenol A degradation.
Journal of hazardous materials (2014-02-22)
Li-Fen Chiang, Ruey-An Doong
ABSTRACT
In this study, the microwave-assisted sol-gel method and chemical reduction were used to synthesize Cu-TiO2 nanorod composites for enhanced photocatalytic degradation of bisphenol A (BPA) in the presence of UV and visible lights. The electron microscopic images showed that the Cu nanoparticles at 4.5±0.1nm were well-deposited onto the surface of TiO2 nanorods after chemical reduction of Cu ions by NaBH4. The X-ray diffractometry patterns and X-ray photoelectron spectroscopic results indicated that Cu species on the Cu-TiO2 nanorods were mainly the mixture of Cu2O and Cu(0). The Cu-TiO2 nanorods showed excellent photocatalytic activity toward BPA photodegradation under the irradiation of UV and visible lights. The pseudo-first-order rate constant (kobs) for BPA photodegradation by 7wt% Cu-TiO2 nanorods were 18.4 and 3.8 times higher than those of as-synthesized TiO2 nanorods and Degussa P25 TiO2, respectively, under the UV light irradiation. In addition, the kobs for BPA photodegradation by 7wt% Cu-TiO2 nanorods increased by a factor of 5.8 when compared with that of Degussa P25 TiO2 under the irradiation of 460±40nm visible light. Results obtained in this study clearly demonstrate the feasibility of using one-dimensional Cu-TiO2 nanorods for photocatalytic degradation of BPA and other pharmaceutical and personal care products in water and wastewater treatment plants.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
VenPure® SF, powder
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Benzyl alcohol, European Pharmacopoeia (EP) Reference Standard
Oleic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Benzyl alcohol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Benzyl alcohol, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzyl alcohol, ACS reagent, ≥99.0%
Sigma-Aldrich
Benzyl alcohol, puriss., meets analytical specification of Ph. Eur., BP, NF, 99-100.5% (GC)
Supelco
Benzyl alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Bisphenol A, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Oleic acid, Selectophore, ≥99%
Sigma-Aldrich
Tetramethylammonium hydroxide pentahydrate, ≥97%
Supelco
Bisphenol A, ≥99%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Benzyl alcohol, anhydrous, 99.8%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%