Merck
Anticancer effects of baicalein on hepatocellular carcinoma cells.
Phytotherapy research : PTR (2014-03-07)
Yi-Hu Zheng, Li-Hui Yin, Tan Hooi Min Grahn, Ai-Fang Ye, Yan-Rong Zhao, Qi-Yu Zhang
ABSTRACT
The therapeutic potential of baicalein against hepatoma cells was evaluated in vitro and in vivo. In cell viability assays, baicalein showed significant cytotoxicity against the hepatocellular carcinoma cell lines H22, Bel-7404, and Hep G2 and moderate cytotoxicity against immortalized human hepatocytes. Baicalein induced G0/G1-phase arrest in hepatocellular carcinoma cells, inhibited AKT, and promoted the degradation of β-catenin and cyclin D1 without activation of GSK-3β. Furthermore, baicalein significantly inhibited H22 xenograft tumor growth without causing obvious adverse effects on weight or liver and spleen weight indexes in ICR mice. Immunohistochemical analysis showed that the inhibition of tumor growth in baicalein-treated mice was associated with decreased AKT, β-catenin, and cyclin D1 expression ex vivo. Our data indicate that baicalein might regulate cyclin D1 transcription via a β-catenin-dependent mechanism, leading to cell cycle arrest at G0/G1 phase and impaired cancer cell proliferation. These results suggest that baicalein is a potential candidate for the treatment of hepatocellular carcinoma.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, absolute, sales not in Germany, ≥99.8% (vol.)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Supelco
Ethanol, standard for GC
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Cyclophosphamide monohydrate, bulk package
Sigma-Aldrich
Cyclophosphamide monohydrate, ISOPAC®
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Baicalein, 98%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications