Merck
  • Quality assessment of the blue mussel (Mytilus edulis): comparison between commercial and wild types.
Quality assessment of the blue mussel (Mytilus edulis): comparison between commercial and wild types.
Marine pollution bulletin (2014-06-28)
B De Witte, L Devriese, K Bekaert, S Hoffman, G Vandermeersch, K Cooreman, J Robbens
ABSTRACT
This study compared species identity, microplastics, chemical and microbial contamination between consumption mussels and wild type mussels, collected at Belgian department stores and Belgian groynes and quaysides, respectively. Species identification based on genetic analysis showed a high number of Mytilus (M.) edulis compared to M. galloprovincialis and M. edulis/galloprovincialis hybrid mussels. The number of total microplastics varied from 2.6 to 5.1 fibres/10 g of mussel. A higher prevalence of orange fibres at quaysides is related to fisheries activities. Chemical contamination of polycyclic aromatic hydrocarbons and polychlorobiphenyls could be related to industrial activities and water turbidity, with maximum concentrations at the quayside of port Zeebrugge. The inverse was noted for Escherichia coli contamination, which was relatively low at Zeebrugge quayside with a total count of 3.9 × 10(2)CFU/100 g tissue, due to limited agricultural effluents. Results of this complementary analysis stress the importance of integrated monitoring and quality assessment.
Sigma-Aldrich
2-Methylheptane, 98%
Sigma-Aldrich
Perchloric acid, ACS reagent, 60%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Supelco
Perchloric acid, 0.01 M HClO4 in water (0.01N), eluent for IC
Sigma-Aldrich
Perchloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 70.0-72.0%
Sigma-Aldrich
Perchloric acid, 70%, 99.999% trace metals basis
Sigma-Aldrich
Perchloric acid, ACS reagent, 70%
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Supelco
Hexane, analytical standard
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Nitric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥65%
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Nitric acid, puriss. p.a., ≥65% (T)
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Perchloric acid, puriss. p.a., ACS reagent, packed in coated, shock- and leak-protected glass bottle, ≥60% (T)
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous