Merck
  • Dual-targeted polyplexes based on sequence-defined peptide-PEG-oligoamino amides.
Dual-targeted polyplexes based on sequence-defined peptide-PEG-oligoamino amides.
Journal of pharmaceutical sciences (2014-10-01)
Petra Kos, Ulrich Lächelt, Dongsheng He, Yu Nie, Zhongwei Gu, Ernst Wagner
ABSTRACT
For active cell targeting, viruses frequently capitalize on dual-receptor binding. With the intention to mimic this natural process, a dual peptide-based approach for targeting cancer cells was evaluated. For this purpose, sequence-defined pDNA binding oligo (ethane amino) amides containing a PEG chain with a peptidic targeting ligand at its distal end were applied. Integrin receptor-directed cyclic peptide cRGDfk, transferrin receptor-addressing peptide B6, and epidermal growth factor receptor-targeting peptide GE11 were used in the study in DU145 prostate cancer cells that express all three receptors. Dual-receptor targeted pDNA polyplexes were designed by combining two of the above ligands at various ratios, in order to find an optimal ligand combination. Two polycation/pDNA ratios of nitrogen/phosphate (N/P) 6 and 12 were tested. Dual targeting effects were most pronounced at the lower N/P ratio and found for all three combinations. Cell binding studies and pDNA transfections revealed GE11 plus B6 as the most potent combination. In general, a good correlation of cell binding with gene transfer was observed. Interestingly, GE11 peptide-based polyplexes-mediated bimodal cell association profiles. In contrast, B6 ligand, cRGD ligand, and dual-targeted polyplexes triggered more homogenous monomodal cell binding characteristics.
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
2,2′-(Ethylenedioxy)diethanethiol, 95%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Triisopropylsilane, 98%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
2-Chlorotrityl chloride, ≥97.0% (AT)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Supelco
Trifluoroacetic acid, analytical standard
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Supelco
N,N-Dimethylformamide, analytical standard