Merck
  • Ex vivo correlation of the permeability of metoprolol across human and porcine buccal mucosa.
Ex vivo correlation of the permeability of metoprolol across human and porcine buccal mucosa.
Journal of pharmaceutical sciences (2014-05-16)
Emil Meng-Lund, Eva Marxen, Anne Marie L Pedersen, Anette Müllertz, Birgitte Hyrup, Rene Holm, Jette Jacobsen
ABSTRACT
The pH partition theory proposes a correlation between fraction of unionized drug substance and permeability. The aim of this study was to compare the permeability of metoprolol and mannitol in ex vivo human and porcine buccal mucosa models at varying pH to validate whether the porcine permeability model is predictive for human buccal absorption. Human (n = 9-10) and porcine (n = 6-7) buccal mucosa were mounted in a modified Ussing chamber, and the kinetics of metoprolol and mannitol transport was assessed for a period of 5.5 h with the pH values of donor medium set at 7.4, 8.5, and 9.0. In addition, hematoxylin-eosin and Alcian blue-van Gieson were used as tissue stains to evaluate the histology and the presence of acidic polysaccharides (e.g., mucins), respectively. The permeability of metoprolol was decreased in human buccal mucosa by almost twofold when compared with porcine buccal mucosa with a positive correlation (r(2) = 0.96) between the permeability assessed in porcine and human buccal mucosa. There was no change in the degree of either epithelial swelling or desquamation when treating with the pH 9.0 donor medium for 5.5 h. These data suggest that buccal mucosa from pigs can be used to predict human buccal absorption.
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Boric acid-11B, ≥99 atom % 11B
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O