Merck
  • Toxic effects of individual and combined effects of BTEX on Euglena gracilis.
Toxic effects of individual and combined effects of BTEX on Euglena gracilis.
Journal of hazardous materials (2014-12-03)
Cheng Peng, Jong-Wha Lee, Homa Teimouri Sichani, Jack C Ng
ABSTRACT
BTEX is a group of volatile organic compounds consisting of benzene, toluene, ethylbenzene and xylenes. Environmental contamination of BTEX can occur in the groundwater with their effects on the aquatic organisms and ecosystem being sparsely studied. The aim of this study was to evaluate the toxic effects of individual and mixed BTEX on Euglena gracilis (E. gracilis). We examined the growth rate, morphological changes and chlorophyll contents in E. gracilis Z and its mutant SMZ cells treated with single and mixture of BTEX. BTEX induced morphological change, formation of lipofuscin, and decreased chlorophyll content of E. gracilis Z in a dose response manner. The toxicity of individual BTEX on cell growth and chlorophyll inhibition is in the order of xylenes>ethylbenzene>toluene>benzene. SMZ was found more sensitive to BTEX than Z at much lower concentrations between 0.005 and 5 μM. The combined effect of mixed BTEX on chlorophyll contents was shown to be concentration addition (CA). Results from this study suggested that E. gracilis could be a suitable model for monitoring BTEX in the groundwater and predicting the combined effects on aqueous ecosystem.
Product Description
Supelco
m-Xylene, analytical standard
Sigma-Aldrich
m-Xylene, anhydrous, ≥99%
Sigma-Aldrich
Rhein, technical grade
Supelco
m-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
m-Xylene, ReagentPlus®, 99%
Sigma-Aldrich
m-Xylene, puriss. p.a., ≥99.0% (GC)
Supelco
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Rhein
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Ethylbenzene, anhydrous, 99.8%
Supelco
Toluene, analytical standard
Supelco
Ethylbenzene, analytical standard
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Supelco
o-Xylene, analytical standard
Supelco
Benzene, analytical standard
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
o-Xylene, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Supelco
Ethylbenzene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
o-Xylene, suitable for HPLC, 98%
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Benzene, suitable for HPLC, ≥99.9%
Supelco
Benzene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzene, ACS reagent, ≥99.0%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%