Merck
  • Rearrangement of quinoxalin-2-ones when exposed to enamines generated in situ from ketones and ammonium acetate: method for the synthesis of 1-(pyrrolyl)benzimidazolones.
Rearrangement of quinoxalin-2-ones when exposed to enamines generated in situ from ketones and ammonium acetate: method for the synthesis of 1-(pyrrolyl)benzimidazolones.
The Journal of organic chemistry (2014-12-17)
Vakhid A Mamedov, Nataliya A Zhukova, Tat'yana N Beschastnova, Victor V Syakaev, Dmitry B Krivolapov, Ekaterina V Mironova, Anastasiya I Zamaletdinova, Il'dar Kh Rizvanov, Shamil K Latypov
ABSTRACT
The reaction of 3-benzoylquinoxalin-2(1H)-ones with enamines (generated in situ from ammonium acetate and the corresponding methylaryl(hetaryl)ketones) proceeds smoothly to give the corresponding substituted 1-(pyrrolyl)benzimidazolone derivatives in moderate yields through the novel rearrangement of 3-benzoylquinoxalin-2(1H)-ones involving a dual cleavage of the C3═N4 and C2-C3 bonds under mild conditions.
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Supelco
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
4-Nitroaniline, analytical standard
Sigma-Aldrich
4-Nitroaniline, ≥99%