Merck
  • Ketoprofen enantioseparation with a Cinchona alkaloid based stationary phase: enantiorecognition mechanism and release studies.
Ketoprofen enantioseparation with a Cinchona alkaloid based stationary phase: enantiorecognition mechanism and release studies.
Journal of separation science (2014-07-22)
Roccaldo Sardella, Antonella Lisanti, Andrea Carotti, Paolo Blasi, Wolfgang Lindner, Benedetto Natalini
ABSTRACT
With the present contribution, we demonstrate that the baseline separation of ketoprofen enantiomers can be successfully achieved (α = 1.09; R(S) = 1.60) in the reversed-phase mode of elution with a commercially available anion-exchange-based chiral stationary phase, incorporating the quinine 2,6-diisopropylphenyl carbamate derivative as the enantioresolving unit. Focused modification of the eluent composition indicated a stereoselective role of hydrophobic and π-π interactions between the selector and selectand units, besides the prime ionic intermolecular interaction. The mechanistic hypotheses based on the chromatographic data were confirmed by in silico molecular dynamic simulations, which allowed us to establish the network of selector-selectand interactions underlying the stereorecognition process at a molecular level. The validated method was successfully used to evaluate the drug content and release profile of ketoprofen-loaded polymeric film, showing drug homogeneous distribution into the film and no preferential interactions between the polymer and one of the enantiomers, with the racemate released at each time point.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride solution, 0.85%
SAFC
Sodium chloride solution, 5 M
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Ammonium acetate, BioUltra, ≥99.0%
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%