Merck
  • Continuous syntheses of Pd@Pt and Cu@Ag core-shell nanoparticles using microwave-assisted core particle formation coupled with galvanic metal displacement.
Continuous syntheses of Pd@Pt and Cu@Ag core-shell nanoparticles using microwave-assisted core particle formation coupled with galvanic metal displacement.
Nanoscale (2014-06-21)
Masato Miyakawa, Norihito Hiyoshi, Masateru Nishioka, Hidekazu Koda, Koichi Sato, Akira Miyazawa, Toshishige M Suzuki
ABSTRACT
Continuous synthesis of Pd@Pt and Cu@Ag core-shell nanoparticles was performed using flow processes including microwave-assisted Pd (or Cu) core-nanoparticle formation followed by galvanic displacement with a Pt (or Ag) shell. The core-shell structure and the nanoparticle size were confirmed using high-angle annular dark-field scanning transmission electron microscopy (HAADF-STEM) observation and EDS elemental mapping. The Pd@Pt nanoparticles with a particle size of 6.5 ± 0.6 nm and a Pt shell thickness of ca. 0.25 nm were synthesized with appreciably high Pd concentration (Pd 100 mM). This shell thickness corresponds to one atomic layer thickness of Pt encapsulating the Pd core metal. The particle size of core Pd was controlled by tuning the initial concentrations of Na2[PdCl4] and PVP. Core-shell Cu@Ag nanoparticles with a particle size of 90 ± 35 nm and an Ag shell thickness of ca. 3.5 nm were obtained using similar sequential reactions. Oxidation of the Cu core was suppressed by the coating of Cu nanoparticles with the Ag shell.
Product Description
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Silver nitrate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
USP
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethylene glycol solution, suitable for NMR (reference standard), 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Supelco
Ethylene glycol, analytical standard
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Silver nitrate, ≥99.999% trace metals basis
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Sigma-Aldrich
Silver nitrate, meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Silver nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%