Merck
  • Pro1170 Ala polymorphism in HER2-neu is associated with risk of trastuzumab cardiotoxicity.
Pro1170 Ala polymorphism in HER2-neu is associated with risk of trastuzumab cardiotoxicity.
BMC cancer (2015-04-18)
Sasha E Stanton, Maureen M Ward, Paul Christos, Rachel Sanford, Christina Lam, Marta V Cobham, Diana Donovan, Ronald J Scheff, Tessa Cigler, Anne Moore, Linda T Vahdat, Maureen E Lane, Ellen Chuang
ABSTRACT
Variations in single nucleotide polymorphisms (SNPs) have been associated with enhanced drug efficacy and toxicity in cancer therapy. SNP variations in the ErbB2 gene have been identified that alter the protein sequence of the HER2-neu protein, but how these polymorphisms affect prognosis and response to HER2 targeted therapy is unknown. We examined eleven ErbB2 SNPs that alter the HER2-neu amino acid sequence to determine whether any of these particular polymorphisms were associated with increased trastuzumab cardiotoxicity in a case-control study. 140 subjects were enrolled from a single institution under Weill Cornell Medical College IRB protocol #0804009734. Patients were eligible if they had histologically or cytologically proven HER2-neu positive breast cancer and more than 3 months of trastuzumab therapy. Cases had either symptomatic CHF or a decline in LVEF of 15% (or if the LVEF <55%, a decline in LVEF of 10%) that resulted in at least temporary discontinuation of trastuzumab, whereas controls had no decline in their LVEF. Eleven ErbB2 single gene SNPs that resulted in an alteration in the HER2-neu protein amino acid sequence were studied. Single gene SNP analysis was carried out using SNP genotyping assays from genomic DNA obtained from peripheral blood or buccal swab. Only two of the ErbB2 SNPs (Ile 655 Val and Pro 1170 Ala) were found to have variation. There was no association between codon 665 and cardiotoxicity; however the proline variant of amino acid 1170 was more likely than the alanine variant to be found in cases with trastuzumab cardiotoxicity (35% of case patients as compared to 17% of controls, p = 0.04). This association remained significant in multivariable analysis taking into account age, race, and history of hypertension (adjusted OR = 2.60, 95% CI = 1.02, 6.62, p = 0.046). The Her2/neu Pro 1170 Ala polymorphism can be used to identify a subset of patients who are at increased risk of cardiotoxicity from trastuzumab therapy. Her2/neu single nucleotide polymorphisms may be useful in conjunction with other biomarkers to risk stratify patients in order to optimize clinical management.
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard