Merck
  • Nanoparticles based on a hydrophilic polyester with a sheddable PEG coating for protein delivery.
Nanoparticles based on a hydrophilic polyester with a sheddable PEG coating for protein delivery.
Pharmaceutical research (2014-03-15)
Neda Samadi, Mies J van Steenbergen, Joep B van den Dikkenberg, Tina Vermonden, Cornelus F van Nostrum, Maryam Amidi, Wim E Hennink
ABSTRACT
To investigate the effect of polyethylene glycol (PEG) in nanoparticles based on blends of hydroxylated aliphatic polyester, poly(D,L-lactic-co-glycolic-co-hydroxymethyl glycolic acid) (PLGHMGA) and PEG-PLGHMGA block copolymers on their degradation and release behavior. Protein-loaded nanoparticles were prepared with blends of varying ratios of PEG-PLGHMGA (molecular weight of PEG 2,000 and 5,000 Da) and PLGHMGA, by a double emulsion method with or without using poly(vinyl alcohol) (PVA) as surfactant. Bovine serum albumin and lysozyme were used as model proteins. PEGylated particles prepared without PVA had a zeta potential ranging from ~ -3 to ~-35 mV and size ranging from ~200 to ~600 nm that were significantly dependent on the content and type of PEG-block copolymer. The encapsulation efficiency of the two proteins however was very low (<30%) and the particles rapidly released their content in a few days. In contrast, all formulations prepared with PVA showed almost similar particle properties (size: ~250 nm, zeta potential: ~-1 mV), while loading efficiency for both model proteins was rather high (80-90%). Unexpectedly, independent of the type of formulation, the nanoparticles had nearly the same release and degradation characteristics. NMR analysis showed almost a complete removal of PEG in 5 days which explains these marginal differences. Protein release and particle degradation are not substantially influenced by the content of PEG, likely because of the fast shedding of the PEG blocks. These PEG shedding particles are interesting system for intracellular delivery of drugs.
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Supelco
Benzyl alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium phosphate dibasic, purum p.a., anhydrous, ≥98.0% (T)
Supelco
Toluene, analytical standard
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
4-(Dimethylamino)pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzyl alcohol, anhydrous, 99.8%
Sigma-Aldrich
Benzyl alcohol, ≥99%, FCC, FG
Sigma-Aldrich
Benzyl alcohol, natural, ≥98%, FG
Supelco
Benzyl alcohol, analytical standard
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, Molecular Biology, anhydrous, ≥98%
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
4-(Dimethylamino)pyridine, purum, ≥98.0% (NT)
Sigma-Aldrich
Sodium phosphate dibasic, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, Molecular Biology, ≥98.5% (titration)
USP
Benzyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Benzyl alcohol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Benzyl alcohol, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, free-flowing, Redi-Dri