Merck
  • Species distribution and susceptibility profile to fluconazole, voriconazole and MXP-4509 of 551 clinical yeast isolates from a Romanian multi-centre study.
Species distribution and susceptibility profile to fluconazole, voriconazole and MXP-4509 of 551 clinical yeast isolates from a Romanian multi-centre study.
European journal of clinical microbiology & infectious diseases : official publication of the European Society of Clinical Microbiology (2014-09-17)
B Minea, V Nastasa, R F Moraru, A Kolecka, M M Flonta, I Marincu, A Man, F Toma, M Lupse, B Doroftei, N Marangoci, M Pinteala, T Boekhout, M Mares
ABSTRACT
This is the first multi-centre study regarding yeast infections in Romania. The aim was to determine the aetiological spectrum and susceptibility pattern to fluconazole, voriconazole and the novel compound MXP-4509. The 551 isolates were identified using routine laboratory methods, matrix-assisted laser desorption ionisation time-of-flight mass spectrometry (MALDI-TOF MS) and DNA sequence analysis. Susceptibility testing was performed using the European Committee for Antimicrobial Susceptibility Testing (EUCAST) method and breakpoints. The yeasts originated from superficial infections (SUP, 51.5 %), bloodstream infections (BSI, 31.6 %) and deep-seated infections (DEEP, 16.9 %), from patients of all ages. Nine genera and 30 species were identified. The 20 Candida species accounted for 94.6 % of all isolates. C. albicans was the overall leading pathogen (50.5 %). Lodderomyces elongisporus is reported for the first time as a fungaemia cause in Europe. C. glabrata and Saccharomyces cerevisiae, as well as the non-Candida spp. and non-albicans Candida spp. groups, showed decreased fluconazole susceptibility (<75 %). The overall fluconazole resistance was 10.2 %. C. krusei accounted for 27 of the 56 fluconazole-resistant isolates. The overall voriconazole resistance was 2.5 % and was due mainly to C. glabrata and C. tropicalis isolates. Fluconazole resistance rates for the three categories of infection were similar to the overall value; voriconazole resistance rates differed: 4 % for BSI, 3.2 % for DEEP and 1.4 % for SUP. The antifungal activity of MXP-4509 was superior to voriconazole against C. glabrata and many fluconazole-resistant isolates. There was a large percentage of non-albicans Candida isolates. A large part of the high fluconazole resistance was not acquired but intrinsic, resulting from the high percentage of C. krusei.
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, suitable for MALDI-TOF MS
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 99%
Supelco
α-Cyano-4-hydroxycinnamic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 97%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, ≥98% (TLC), powder
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, suitable for matrix substance for MALDI-MS, Ultra pure
Fluconazole for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Fluconazole, ≥98% (HPLC), powder
Sigma-Aldrich
Voriconazole, ≥98% (HPLC)
USP
Fluconazole, United States Pharmacopeia (USP) Reference Standard
Fluconazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Voriconazole, VETRANAL®, analytical standard
Voriconazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Fluconazole, Pharmaceutical Secondary Standard; Certified Reference Material