Merck
  • Effects of N source concentration and NH4(+)/NO3(-) ratio on phenylethanoid glycoside pattern in tissue cultures of Plantago lanceolata L.: a metabolomics driven full-factorial experiment with LC-ESI-MS(3.).
Effects of N source concentration and NH4(+)/NO3(-) ratio on phenylethanoid glycoside pattern in tissue cultures of Plantago lanceolata L.: a metabolomics driven full-factorial experiment with LC-ESI-MS(3.).
Phytochemistry (2014-08-02)
Sándor Gonda, Attila Kiss-Szikszai, Zsolt Szűcs, Csaba Máthé, Gábor Vasas
ABSTRACT
Tissue cultures of a medicinal plant, Plantago lanceolata L. were screened for phenylethanoid glycosides (PGs) and other natural products (NPs) with LC-ESI-MS(3). The effects of N source concentration and NH4(+)/NO3(-) ratio were evaluated in a full-factorial (FF) experiment. N concentrations of 10, 20, 40 and 60mM, and NH4(+)/NO3(-) ratios of 0, 0.11, 0.20 and 0.33 (ratio of NH4(+) in total N source) were tested. Several peaks could be identified as PGs, of which, 16 could be putatively identified from the MS/MS/MS spectra. N source concentration and NH4(+)/NO3(-) ratio had significant effects on the metabolome, their effects on individual PGs were different despite these metabolites were of the same biosynthethic class. Chief PGs were plantamajoside and acteoside (verbascoside), their highest concentrations were 3.54±0.83% and 1.30±0.40% of dry weight, on media 10(0.33) and 40(0.33), respectively. NH4(+)/NO3(-) ratio and N source concentration effects were examined on a set of 89 NPs. For most NPs, high increases in abundance were observed compared to Murashige-Skoog medium. Abundances of 42 and 10 NPs were significantly influenced by the N source concentration and the NH4(+)/NO3(-) ratio, respectively. Optimal media for production of different NP clusters were 10(0), 10(0.11) and 40(0.33). Interaction was observed between NH4(+)/NO3(-) ratio and N source concentration for many NPs. It was shown in simulated experiments, that one-factor at a time (OFAT) experimental designs lead to sub-optimal media compositions for production of many NPs, and alternative experimental designs (e.g. FF) should be preferred when optimizing medium N source for optimal yield of NPs. If using OFAT, the N source concentration is to be optimized first, followed by NH4(+)/NO3(-) ratio, as this reduces the likeliness of suboptimal yield results.
Product Description
Supelco
Pyridoxine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Thiamine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Pyridoxine hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Thiamine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Supelco
2,4-D, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
2,4-D, PESTANAL®, analytical standard
Sigma-Aldrich
Thiamine hydrochloride, ≥98%, FCC, FG
Supelco
Thiamine Hydrochloride (B1), analytical standard
USP
Pyridoxine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Thiamine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Thiamine hydrochloride, tested according to Ph. Eur.
Sigma-Aldrich
Thiamine hydrochloride, reagent grade, ≥99% (HPLC)
Sigma-Aldrich
Pyridoxine hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Pyridoxine hydrochloride, meets USP testing specifications
Sigma-Aldrich
Pyridoxine hydrochloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Thiamine hydrochloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
Supelco
Pyridoxine Hydrochloride (B6), analytical standard
Sigma-Aldrich
Thiamine hydrochloride, meets USP testing specifications
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
2,4-D, PESTANAL®, analytical standard
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)