Merck
  • Silencing of PINK1 inhibits insulin-like growth factor-1-mediated receptor activation and neuronal survival.
Silencing of PINK1 inhibits insulin-like growth factor-1-mediated receptor activation and neuronal survival.
Journal of molecular neuroscience : MN (2014-12-24)
María J Contreras-Zárate, Andrea Niño, Liliana Rojas, Humberto Arboleda, Gonzalo Arboleda
ABSTRACT
The etiology of Parkinson's disease remains unknown. Mutations in PINK1 have provided an understanding of the molecular mechanisms of this pathology. PINK1 and Parkin are important in the dismissal of dysfunctional mitochondria. However, the role of PINK1 in the control of neuronal survival pathways is not clear. To determine the role of PINK1 in the control of the phosphatidyl inositol 3-kinase (PI3K)/Akt pathway mediated by insulin-like grow factor type 1 (IGF-1), we use a model of mesencephalic neurons (CAD cells), which were transfected with lentiviral PINK1 shRNA or control shRNA constructs. Silencing of PINK1 was determined by RT-PCR and immunoblotting; cell viability was analyzed by 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) and lactate dehydrogenase (LDH) assays; proteins of the PI3K/Akt signaling pathway were tested by immunoblotting and IGF-1 receptor, and mitochondria were examined using fluorescence microscopy. PINK1 shRNA-transfected cells showed a reduction in cell survival compared to control shRNA cells. Exposure to IGF-1 induced a rapid and high increase in the phosphorylation level of IGF-1 receptor in control shRNA-transfected cells; however, silencing of PINK1 decreases phosphorylation level of IGF-1 receptor and downstream target proteins such as Akt, GSK3-beta, IRS-1, and hexokinase. Our results further suggest that PINK1 may be regulating the PI3K/Akt neuronal survival pathway through tyrosine kinase receptors such as IGF-1 receptor.
Product Description
Supelco
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
L-Glutamine