Merck
  • Pleural innate response activator B cells protect against pneumonia via a GM-CSF-IgM axis.
Pleural innate response activator B cells protect against pneumonia via a GM-CSF-IgM axis.
The Journal of experimental medicine (2014-05-14)
Georg F Weber, Benjamin G Chousterman, Ingo Hilgendorf, Clinton S Robbins, Igor Theurl, Louisa M S Gerhardt, Yoshiko Iwamoto, Tam D Quach, Muhammad Ali, John W Chen, Thomas L Rothstein, Matthias Nahrendorf, Ralph Weissleder, Filip K Swirski
ABSTRACT
Pneumonia is a major cause of mortality worldwide and a serious problem in critical care medicine, but the immunophysiological processes that confer either protection or morbidity are not completely understood. We show that in response to lung infection, B1a B cells migrate from the pleural space to the lung parenchyma to secrete polyreactive emergency immunoglobulin M (IgM). The process requires innate response activator (IRA) B cells, a transitional B1a-derived inflammatory subset which controls IgM production via autocrine granulocyte/macrophage colony-stimulating factor (GM-CSF) signaling. The strategic location of these cells, coupled with the capacity to produce GM-CSF-dependent IgM, ensures effective early frontline defense against bacteria invading the lungs. The study describes a previously unrecognized GM-CSF-IgM axis and positions IRA B cells as orchestrators of protective IgM immunity.
Sigma-Aldrich
2-Methylbutane, anhydrous, ≥99%
Supelco
2-Methylbutane, analytical standard
Sigma-Aldrich
3-Amino-9-ethylcarbazole, tablet
Sigma-Aldrich
3-Amino-9-ethylcarbazole, ≥95% (HPLC), powder
Sigma-Aldrich
2-Methylbutane, suitable for HPLC, ≥99.5%
Sigma-Aldrich
2-Methylbutane, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Methylbutane, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Methylbutane, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material