Merck
  • Cre-loxP-based system for removal and reuse of selection markers in Ashbya gossypii targeted engineering.
Cre-loxP-based system for removal and reuse of selection markers in Ashbya gossypii targeted engineering.
Fungal genetics and biology : FG & B (2014-05-06)
Tatiana Q Aguiar, Cláudia Dinis, Lucília Domingues
ABSTRACT
The filamentous ascomycete Ashbya gossypii is amenable to genetic manipulation and is an excellent model system for studying eukaryotic cell biology. However, the number of selection markers in current use for both targeted gene integration and disruption in this fungus are very limited. Therefore, the Cre-loxP recombination system was adapted for use in A. gossypii and its effectiveness in recycling marker genes was demonstrated by constructing both single and double deleted Agura3 and Agade1 auxotrophic strains free of exogenous markers. In spite of its wide use in other organisms, including other Ascomycete fungi, this is the first report describing Cre-loxP-based methodology for A. gossypii, opening new perspectives for targeted engineering of this fungus with several promising biotechnological applications [corrected].
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Ampicillin, anhydrous, European Pharmacopoeia (EP) Reference Standard
USP
Ampicillin, United States Pharmacopeia (USP) Reference Standard
Supelco
Ampicillin, analytical standard
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Adenine, BioReagent, suitable for cell culture
Sigma-Aldrich
Adenine, BioReagent, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Adenine, ≥99%
Adenine, European Pharmacopoeia (EP) Reference Standard
Supelco
Adenine, Pharmaceutical Secondary Standard; Certified Reference Material