Merck
  • Simultaneous determination of 16 phenolic constituents in Spatholobi Caulis by high performance liquid chromatography/electrospray ionization triple quadrupole mass spectrometry.
Simultaneous determination of 16 phenolic constituents in Spatholobi Caulis by high performance liquid chromatography/electrospray ionization triple quadrupole mass spectrometry.
Journal of pharmaceutical and biomedical analysis (2014-09-30)
Yu Zhang, Long Guo, Li Duan, Xin Dong, Ping Zhou, E-Hu Liu, Ping Li
ABSTRACT
Spatholobi Caulis, the vine stem of Spatholobus suberectus Dunn, has been widely used in traditional Chinese and folk medicines for treatment of irregular menstruation, blood deficiency and rheumatalgia in clinic. In this study, an accurate and reliable high performance liquid chromatography/electrospray ionization triple quadrupole mass spectrometry (HPLC-MS/MS) method was established for simultaneous determination of 16 phenolic bioactive constituents, including five flavanols, seven isoflavonoids, three flavanones and one chalcone in Spatholobi Caulis. The method validation results exhibited that the developed method had desirable specificity, linearity, precision and accuracy. The quantitative analysis results showed that flavanols were the abundant constituents in Spatholobi Caulis. Moreover, principal component analysis (PCA) was performed to assess the quality variation of samples collected from different regions in China. The PCA results indicated the quantitative analysis based on HPLC-MS/MS is a feasible method for quality assessment and control of Spatholobi Caulis.
Sigma-Aldrich
5,7-Dimethoxycoumarin, 98%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard