Merck
  • Characterization of in vitro metabolites of JWH-018, JWH-073 and their 4-methyl derivatives, markers of the abuse of these synthetic cannabinoids.
Characterization of in vitro metabolites of JWH-018, JWH-073 and their 4-methyl derivatives, markers of the abuse of these synthetic cannabinoids.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-03-25)
Veniero Gambaro, Sebastiano Arnoldi, Stefania Bellucci, Eleonora Casagni, Lucia Dell'Acqua, Laura Fumagalli, Marco Pallavicini, Gabriella Roda, Chiara Rusconi, Ermanno Valoti
ABSTRACT
In vitro incubation with human liver microsomes of JWH-018, JWH-073, JWH-122 and for the first time 1-butyl-3-(1-(4-methyl)naphthoyl)indole (the 4-methylnaphthoyl analogue of JWH-073) was investigated to identify the principal metabolites of alkylindole synthetic cannabinoids, thus helping the discovering of synthetic cannabinoids abusers. The results obtained showed that the most abundant metabolites were mono-hydroxylated derivatives either on the alkyl chain (ω or ω-1 position) or on the indole (presumably in position 5 or 6) and naphthalene moieties. Moreover the extraction conditions of these derivatives from biological fluids, mainly plasma and urine spiked with commercially available metabolite standards, and the incubation procedure were investigated to obtain a fast, reliable and suitable extraction protocol to detect either the parent drugs or their metabolites by means of GC/MS.
Product Description
Supelco
Sodium carbonate concentrate, 0.1 M Na2CO3 in water, eluent concentrate for IC
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
N,O-Bis(trimethylsilyl)trifluoroacetamide, ≥99%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
Dichloromethane, Selectophore, ≥99.5%
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)