Merck
Pseudomycoicidin, a Class II Lantibiotic from Bacillus pseudomycoides.
Applied and environmental microbiology (2015-03-15)
Shradha Basi-Chipalu, Jasmin Dischinger, Michaele Josten, Christiane Szekat, Annegret Zweynert, Hans-Georg Sahl, Gabriele Bierbaum
ABSTRACT
Lantibiotics are ribosomally synthesized antimicrobial peptides with substantial posttranslational modifications. They are characterized by the unique amino acids lanthionine and methyllanthionine, which are introduced by dehydration of Ser/Thr residues and linkage of the resulting dehydrated amino acids with Cys residues. BLAST searches using the mersacidin biosynthetic enzyme (MrsM) in the NCBI database revealed a new class II lantibiotic gene cluster in Bacillus pseudomycoides DSM 12442. Production of an antimicrobial substance with activity against Gram-positive bacteria was detectable in a cell wash extract of this strain. The substance was partially purified, and mass spectrometric analysis predicted a peptide of 2,786 Da in the active fraction. In order to characterize the putative lantibiotic further, heterologous expression of the predicted biosynthetic genes was performed in Escherichia coli. Coexpression of the prepeptide (PseA) along with the corresponding modification enzyme (PseM) resulted in the production of a modified peptide with the corresponding mass, carrying four out of eight possible dehydrations and supporting the presence of four thioether and one disulfide bridge. After the proteolytic removal of the leader, the core peptide exhibited antimicrobial activity. In conclusion, pseudomycoicidin is a novel lantibiotic with antimicrobial activity that was heterologously produced in E. coli.
Sigma-Aldrich
3-Indoleacetic acid, 98%
Sigma-Aldrich
3-Indoleacetic acid, suitable for plant cell culture, crystalline
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
Supelco
3-Indoleacetic acid, PESTANAL®, analytical standard
Sigma-Aldrich
Nitrilotriacetic acid trisodium salt, Sigma Grade, ≥98%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Nitrilotriacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Nitrilotriacetic acid, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Nitrilotriacetic acid, Sigma Grade, ≥99%
Supelco
Nitrilotriacetic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Ultrapure Acetonitrile
USP
Levothyroxine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Acetonitrile, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
Acetonitrile, ≥99.8% (GC)
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)