Merck
  • Effect of prolactin-induced protein on human skin: new insight into the digestive action of this aspartic peptidase on the stratum corneum and its induction of keratinocyte proliferation.
Effect of prolactin-induced protein on human skin: new insight into the digestive action of this aspartic peptidase on the stratum corneum and its induction of keratinocyte proliferation.
The Journal of investigative dermatology (2014-10-15)
Shuji Sugiura, Misao Tazuke, Shoichi Ueno, Yasuo Sugiura, Ikuo Kato, Yoshimitsu Miyahira, Yutaka Yamamoto, Hiroshi Sato, Jun Udagawa, Masami Uehara, Hisashi Sugiura
ABSTRACT
Human prolactin-induced protein (PIP) is a major protein found in exocrine fluids such as saliva and sweat. Intriguingly, PIP possesses residues (human PIP (hPIP): PIP (29-63)) that display similarity to the aspartic peptidase candidapepsin. Here, we aimed to determine the effect of PIP as a protease on normal skin structure. Using an adhesive tape-stripping technique, we applied hPIP peptide on the corneocytes of normal-appearing facial skin from infants with eczema and healthy infants and then analyzed the morphological structure of corneocytes with Nile Red fluorescence. We also repeatedly applied the hPIP peptide onto the surface of a three-dimensional (3-D) human skin model and then analyzed any changes to the stratum corneum and epidermis using light microscopy and scanning electron microscopy. In both infant groups, a decrease in hydrophobic lipids from the cornified envelope was observed after treatment with hPIP. The peptide hPIP appeared to digest the fine structure of the stratum corneum and induce a proliferation of epidermal keratinocytes within the 3-D human skin model. Our results suggest that aspartic peptidase of PIP found in sweat or saliva deteriorates the skin barrier in a de novo manner, which potentially leads directly to the proliferation of epidermal keratinocytes without any external antigenic factors.
Product Description
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Supelco
2-Mercaptoethanol, derivatization grade (HPLC), LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ACS reagent
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture