Merck
  • Metabolic profiling of plant extracts using direct-injection electrospray ionization mass spectrometry allows for high-throughput phenotypic characterization according to genetic and environmental effects.
Metabolic profiling of plant extracts using direct-injection electrospray ionization mass spectrometry allows for high-throughput phenotypic characterization according to genetic and environmental effects.
Journal of agricultural and food chemistry (2015-01-15)
Martín García-Flores, Sheila Juárez-Colunga, Adrián García-Casarrubias, Samuel Trachsel, Robert Winkler, Axel Tiessen
ABSTRACT
In comparison to the exponential increase of genotyping methods, phenotyping strategies are lagging behind in agricultural sciences. Genetic improvement depends upon the abundance of quantitative phenotypic data and the statistical partitioning of variance into environmental, genetic, and random effects. A metabolic phenotyping strategy was adapted to increase sample throughput while saving reagents, reducing cost, and simplifying data analysis. The chemical profiles of stem extracts from maize plants grown under low nitrogen (LN) or control trial (CT) were analyzed using optimized protocols for direct-injection electrospray ionization mass spectrometry (DIESI-MS). Specific ions significantly decreased or increased because of environmental (LN versus CT) or genotypic effects. Biochemical profiling with DIESI-MS had a superior cost-benefit compared to other standard analytical technologies (e.g., ultraviolet, near-infrared reflectance spectroscopy, high-performance liquid chromatography, and gas chromatography with flame ionization detection) routinely used for plant breeding. The method can be successfully applied in maize, strawberry, coffee, and other crop species.
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Fluorescein (free acid), Dye content 95 %
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ACS reagent
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard