Merck
  • Multiresidue method for the determination of pharmacologically active substances in egg and honey using a continuous solid-phase extraction system and gas chromatography-mass spectrometry.
Multiresidue method for the determination of pharmacologically active substances in egg and honey using a continuous solid-phase extraction system and gas chromatography-mass spectrometry.
Food chemistry (2015-02-24)
Abdelmonaim Azzouz, Evaristo Ballesteros
ABSTRACT
A sensitive, selective, efficient gas chromatography-mass spectrometry method for the simultaneous determination of 22 pharmacologically active substances (antibacterials, nonsteroidal antiinflammatories, antiseptics, antiepileptics, lipid regulators, β-blockers and hormones) in eggs and honey was developed. The sample pretreatment includes precipitation of proteins and lipids with acetonitrile:water (3:2, v/v), centrifugation and continuous solid-phase extraction for cleanup and preconcentration. The proposed method was validated with quite good analytical results including limits of detection of 0.4-3.3 ng/kg for 2g of sample and good linearity (r(2)>0.995) throughout the studied concentration ranges. The recoveries of analytes from real honey and egg samples spiked at concentrations of 15-2,000 ng/kg fell in the range 87-102%, with relative standard deviations from 2.6% to 7.0%. The method was successfully used to determine the target compounds in various types of eggs (hen, quail and duck) and honey samples (flower, forest, acacia, sunflower, clover and pine tree). Two samples of hen eggs bought at supermarkets and one of quail eggs were found to contain florfenicol, pyrimethamine, estrone and 17β-estradiol at levels from 0.095 to 2.7 μg/kg.
Product Description
Supelco
Ibuprofen
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, derivatization grade (GC derivatization), LiChropur, contains 10% TMCS, 98% (excluding TMCS)
Sigma-Aldrich
Chlorotrimethylsilane, puriss., ≥99.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Chlorotrimethylsilane, Wacker Chemie AG, ≥99.0% (GC)
Supelco
Acetone, analytical standard
Sigma-Aldrich
Hexamethyldisiloxane, viscosity 0.65 cSt (25 °C)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Thiamphenicol
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, with 1% trimethylchlorosilane, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Phenylbutazone
Sigma-Aldrich
Mefenamic acid
Sigma-Aldrich
Estrone, ≥99%
USP
Estrone, United States Pharmacopeia (USP) Reference Standard