Merck
  • Generation of stable L. major(+EGFP-LUC) and simultaneous comparison between EGFP and luciferase sensitivity.
Generation of stable L. major(+EGFP-LUC) and simultaneous comparison between EGFP and luciferase sensitivity.
Experimental parasitology (2015-02-01)
Tahereh Taheri, Hana Saberi Nik, Negar Seyed, Fatemeh Doustdari, Mohammad-Hossein Etemadzadeh, Fatemeh Torkashvand, Sima Rafati
ABSTRACT
Because of the lack of an accurate and sensitive tool to evaluate the parasitemia level, treatment or prevention of leishmaniasis remains an important challenge worldwide. To monitor and track leishmanial infection by two parameters in real time, we generated stably transgenic Leishmania that express a bi-reporter protein as fused EGFP and firefly luciferase. Using two reporter genes (egfp-luc) simultaneously increases the experimental sensitivity for detection/diagnosis, and in vitro quantification of parasites as well as real-time infection in mice. Through different specific tools, EGFP and LUC signals from the parasite were detectable and measurable within a mammalian host and promastigotes. Here, the LUC protein provided a higher level of sensitivity than did EGFP, so that infection was detectable at an earlier stage of the disease in the footpad (injection site) and lymph nodes by bioluminescence. These results depicted that: (1) both quantitative reporter genes, EGFP and LUC, could be simultaneously used to detect parasitemia in vitro and in vivo and (2) sensitivity of firefly luciferase was 10-fold higher than that of EGFP in promastigotes.
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Adenosine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
Adenosine, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Adenosine, ≥99%
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine
Supelco
Adenosine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Adenosine, European Pharmacopoeia (EP) Reference Standard