Merck
  • Identification of novel transcription factors in osteoclast differentiation using genome-wide analysis of open chromatin determined by DNase-seq.
Identification of novel transcription factors in osteoclast differentiation using genome-wide analysis of open chromatin determined by DNase-seq.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research (2014-03-29)
Kazuki Inoue, Yuuki Imai
ABSTRACT
Clarification of the mechanisms underlying osteoclast differentiation enables us to understand the physiology of bone metabolism as well as the pathophysiology of bone diseases such as osteoporosis. Recently, it has been reported that epigenetics can determine cell fate and regulate cell type-specific gene expression. However, little is known about epigenetics during osteoclastogenesis. To reveal a part of epigenetics, especially focused on chromatin dynamics, during early osteoclastogenesis and to identify novel transcription factors involved in osteoclastogenesis, we performed a genome-wide analysis of open chromatin during receptor activator of NF-κB ligand (RANKL)-induced osteoclastogenesis using DNase I hypersensitive sites sequencing (DNase-seq). DNase-seq was performed using the extracted nuclei from RAW264 cells treated with or without RANKL for 24 hours, followed by several bioinformatic analyses. DNase I hypersensitive sites (DHSs) were dynamically changed during RANKL-induced osteoclastogenesis and they accumulated in promoter regions. The distributions of DHSs among cis-regulatory DNA regions were identical regardless of RANKL stimulation. Motif discovery analysis successfully identified well-known osteoclastogenic transcription factors including Jun, CREB1, FOS, ATF2, and ATF4, but also novel transcription factors for osteoclastogenesis such as Zscan10, Atf1, Nrf1, and Srebf2. siRNA knockdown of these identified novel transcription factors impaired osteoclastogenesis. Taken together, DNase-seq is a useful tool for comprehension of epigenetics, especially chromatin dynamics during osteoclastogenesis and for identification of novel transcription factors involved in osteoclastogenesis. This study may reveal underlying mechanisms that determine cell type-specific differentiation of bone cells and may lead to investigation of novel therapeutic targets for osteoporosis.
Sigma-Aldrich
MISSION® esiRNA, targeting human SREBF2
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Srebf2
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder