Merck
  • Evaluation of edaravone against radiation-induced oral mucositis in mice.
Evaluation of edaravone against radiation-induced oral mucositis in mice.
Journal of pharmacological sciences (2015-04-04)
Noriko Nakajima, Shinichi Watanabe, Takeshi Kiyoi, Akihiro Tanaka, Katsuya Suemaru, Hiroaki Araki
ABSTRACT
Oral mucositis induced by radiotherapy for cancers of the head and neck reduce the quality of life of patients. However, effective therapeutic agents are lacking. Symptomatic treatment involves local anesthesia and analgesia. We focused on the antioxidant effects of edaravone (3-methyl-1-phenyl-2-pyrazolin-5-one; Radicut(®)). Oral mucositis was induced on the tongue tips of mice using a single dose of X-rays (20 Gy). To evaluate the protective effect of edaravone (30 and 300 mg/kg), administration was carried out 30 min before irradiation. Survival, oral mucositis score, myeloperoxidase activity, and levels of 2-Thiobarbituric acid reactive substances were measured, and all were improved compared with those of control mice. A significant difference was not found in terms of survival due to edaravone. Histopathologic findings also highlighted the beneficial features of edaravone. Edaravone reduced the production of reactive oxygen species. These findings suggest that the protective effect of edaravone against radiation-induced oral mucositis is through an antioxidant effect.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
o-Dianisidine dihydrochloride, ≥95%
Sigma-Aldrich
o-Dianisidine dihydrochloride, tablet, 10 mg substrate per tablet
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
o-Dianisidine dihydrochloride, Suitable for use in glucose determination
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O