Merck
  • Thermosensitive molecularly imprinted polymers on porous carriers: preparation, characterization and properties as novel adsorbents for bisphenol A.
Thermosensitive molecularly imprinted polymers on porous carriers: preparation, characterization and properties as novel adsorbents for bisphenol A.
Talanta (2014-08-28)
Ruichen Dong, Jinhua Li, Hua Xiong, Wenhui Lu, Hailong Peng, Lingxin Chen
ABSTRACT
Thermosensitive molecularly imprinted polymers (T-MIPs) on porous carriers were prepared via the synergy of dual functional monomers of 4-vinylpyridine (VP) and N-isopropylacrylamide (NIPAM), for selective recognition and controlled adsorption and release of bisphenol A (BPA) by the temperature regulation. The porous polymer supporter was synthesized by multistep swelling of polystyrene and then both the NIPAM with temperature responsiveness and the basic monomers of VP were grafted on them in a simple way. The resultant T-MIPs showed high binding capacity, fast kinetics, and the adsorption processes were found to follow Langmuir-Freundlich isotherm and pseudo-second-order kinetic models. The adsorption capacity increased slightly along with the rise of temperature (such as 20°C) under lower critical solution temperature (LCST, 33°C) and decreased fast above LCST (such as 50°C). Subsequently, the T-MIPs were employed as novel adsorbents for selective solid-phase extraction (SPE) of BPA from seawater and yogurt samples. Satisfying recoveries in the range of 94.83-98.47% were obtained with the precision of 3.21% at ambient temperature (20°C). Through 6 adsorption-desorption cycles, the reusable T-MIPs exhibited a good recoverability with the relative standard error within 9.8%. The smart T-MIPs provided great potentials for selective identification, adsorption/release and removal of BPA by simple stimuli responsive regulation.
Product Description
Supelco
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Estrone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzoyl peroxide blend with dicyclohexyl phthalate, suitable for use as a catalyst for electron microscopy. Modified to render it safe in transit.
Sigma-Aldrich
Estrone, ≥99%
Sigma-Aldrich
Dibutyl phthalate, 99%
Supelco
Dibutyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Estrone, United States Pharmacopeia (USP) Reference Standard
Estrone, European Pharmacopoeia (EP) Reference Standard
Dibutyl phthalate, European Pharmacopoeia (EP) Reference Standard
Supelco
Dibutyl phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Luperox® A75FP, Benzoyl peroxide, 75% remainder water, contains 25 wt. % water as stabilizer, 75%
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Methacrylic acid, contains 250 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
4-Vinylpyridine, contains 100 ppm hydroquinone as inhibitor, 95%
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, tested according to Ph. Eur.
Supelco
Toluene, analytical standard
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Estriol, ≥97%
Sigma-Aldrich
Estriol, meets USP testing specifications
Sigma-Aldrich
DBP, 98% (HPLC)
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dichloromethane, analytical standard
Estriol, European Pharmacopoeia (EP) Reference Standard
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, meets analytical specification of Ph. Eur., BP, E339, 98.5-102.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, puriss. p.a., crystallized, ≥99.0% (T)