Merck
  • Characterization of mesenchymal progenitor cells in crown and root pulp from human mesiodentes.
Characterization of mesenchymal progenitor cells in crown and root pulp from human mesiodentes.
Oral diseases (2014-03-13)
M Sato, T Toriumi, N Watanabe, E Watanabe, D Akita, T Mashimo, Y Akiyama, K Isokawa, T Shirakawa, M J Honda
ABSTRACT
Mesiodentes are usually found in the central position of the upper or lower jaw as supernumerary teeth. Here, we obtained 10 mesiodentes and three permanent teeth (PT) and separated the dental pulp (DP) from these into crown and root portions. We then characterized and compared the isolated crown portion-derived cells (crown cells) with root portion-derived cells (root cells) using a range of in vitro assays. Crown cells and root cells were examined for cell surface marker expression, colony-forming unit-fibroblast (CFU-F), cell proliferation, cell cycle characteristics and markers, and osteogenic and adipogenic differentiation. The proportion of CD105-positive cells (CD105(+) cells) in the crown cells vs the root cells varied among the mesiodentes, but not among the PT. When there were more CD105(+) cells in the root cells than in the crown cells, the root cells showed higher CFU-F, proliferation capacity, and osteogenic differentiation capacity. In contrast, when the crown cells contained more CD105(+) cells than the root cells, the crown cells showed the higher CFU-F, proliferation capacity, and osteogenic differentiation capacity. In addition, the sorted CD105(+) cells showed higher CFU-F and proliferation capacity than the sorted CD105(-) cells. These results indicated that proportion of CD105(+) cells is an effective means of characterizing DP-derived cells in mesiodentes.
Product Description
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Calcium, turnings, 99% trace metals basis
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Calcium, pieces, <1 cm, 99%
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1α,25-Dihydroxyvitamin D2 solution, 50 μg/mL in ethanol, 95% (CP)
Sigma-Aldrich
1α,25-Dihydroxyvitamin D2 solution, 100 μg/mL in ethanol, 95% (CP)
Sigma-Aldrich
Calcium, granular, 99%
Sigma-Aldrich
Phenol Red, ACS reagent
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥97.0% (HPLC)
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Toluidine Blue, suitable for microscopy, 8.74% (ZN (THEORY))
SAFC
L-Glutamine
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥99% (HPLC)
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture