Merck
  • Quantitative subcellular study of transferrin receptor-targeted doxorubicin and its metabolite in human breast cancer cells.
Quantitative subcellular study of transferrin receptor-targeted doxorubicin and its metabolite in human breast cancer cells.
European journal of drug metabolism and pharmacokinetics (2013-12-24)
Jinhui Xu, Yuan Sheng, Feifei Xu, Ying Yu, Yun Chen
ABSTRACT
The extended use of doxorubicin (DOX) could be limited due to the emergence of drug resistance and cardiotoxicity associated with its treatment. Conjugates of DOX with transferrin (DOX-TRF) can effectively alleviate these side effects, thereby leading to a better treatment. The effectiveness of DOX-TRF could result from the enhancement of transferrin receptor (TfR)-mediated transportation. However, detailed TfR-mediated DOX delivery has not been fully elucidated thus far, which may rely on the quantitative subcellular study of DOX distribution and metabolism. In this study, an immunoisolation assay was developed to isolate the organelles with high purity, yield and integrity. Using this immunoisolation assay together with liquid chromatography-tandem mass spectrometry (LC/MS/MS), the subcellular distribution profiles of DOX and its main metabolite doxorubicinol (DOXol) in human breast cancer cells MCF-7/WT and MCF-7/ADR were determined and compared after the treatment of DOX and DOX-TRF. As expected, DOX-TRF treated cells have a higher drug accumulation compared to DOX treated cells. DOX-TRF was predominantly cytoplasmic. In addition, TfR-mediated transportation had a significant impact on the transformation of DOX to DOXol in the cells. This study provided the evidence that immunoisolation together with LC/MS/MS is an effective technique in subcellular investigations.
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Mannitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O