Merck
  • Comparative autoradiographic in vitro investigation of melanin concentrating hormone receptor 1 ligands in the central nervous system.
Comparative autoradiographic in vitro investigation of melanin concentrating hormone receptor 1 ligands in the central nervous system.
European journal of pharmacology (2014-05-02)
Cécile Philippe, Daniela Haeusler, Florian Fuchshuber, Helmut Spreitzer, Helmut Viernstein, Marcus Hacker, Wolfgang Wadsak, Markus Mitterhauser
ABSTRACT
The MCHR1 is an interesting pharmacological and pharmaceutical target, due to its involvement in pathologies as diabetes, gut inflammation and adiposity. in vivo PET-studies of the MCHR1 in energy homeostasis and diabetes could be of great value for deeper understanding of endocrinological hormone status and consequential pharmacological interactions. Furthermore, PET-tracers would facilitate compound dose selection of MCHR1 antagonists for treatment. Therefore, we developed two potential PET-tracers, [(11)C]SNAP-7941 and [(18)F]FE@SNAP, for the in vivo visualization of this receptor. Aim of this study was a preclinical in vitro evaluation of both unlabeled ligands. Therefore, a comparative autoradiographic investigation on CNS (coronal rat brain and 4 different human brain regions) and peripheral tissues (rat tongue as target and rat testes as non-target region) was conducted. Competition experiments, using the two radioligands [(125)I]-MCH and [(125)I]-S36057, were performed with selective and specific MCHR1 ligands as PMC-3886, a MCHR1 agonist, SNAP-7941 and FE@SNAP, two MCHR1 antagonists. Additionally, immunohistochemical staining with a specific MCHR1 antibody was performed. Specific binding was found in all tissues known to express the MCHR1 as human and rat CNS and peripheral rat tongue tissue. No specific binding was found in the non-target region of rat testes. MCHR1 antibody staining complemented the outcome of the autoradiographic experiments. The compounds SNAP-7941 and FE@SNAP were generally comparable with PMC-3886. Hence, the in vitro autoradiographic study of the unlabeled compounds SNAP-7941 and FE@SNAP further qualifies the potential of the PET-tracers [(11)C]SNAP-7941 and [(18)F]FE@SNAP as useful MCHR1 PET-tracers.
Sigma-Aldrich
2-Methylbutane, anhydrous, ≥99%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Supelco
2-Methylbutane, analytical standard
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
3-Amino-9-ethylcarbazole, tablet
Sigma-Aldrich
3-Amino-9-ethylcarbazole, ≥95% (HPLC), powder
Sigma-Aldrich
2-Methylbutane, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Methylbutane, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
2-Methylbutane, ReagentPlus®, ≥99%
Captopril for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-Methylbutane, suitable for HPLC, ≥99.5%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Captopril, meets USP testing specifications
Sigma-Aldrich
Captopril, ≥98% (HPLC), powder
Sigma-Aldrich
Calcium chloride
Supelco
Captopril, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Captopril, United States Pharmacopeia (USP) Reference Standard
Captopril, European Pharmacopoeia (EP) Reference Standard