Merck
  • G protein-coupled estrogen receptor (GPER) mediates NSCLC progression induced by 17β-estradiol (E2) and selective agonist G1.
G protein-coupled estrogen receptor (GPER) mediates NSCLC progression induced by 17β-estradiol (E2) and selective agonist G1.
Medical oncology (Northwood, London, England) (2015-03-07)
Changyu Liu, Yongde Liao, Sheng Fan, Hexiao Tang, Zhixiao Jiang, Bo Zhou, Jing Xiong, Sheng Zhou, Man Zou, Jianmiao Wang
ABSTRACT
Estrogen classically drives lung cancer development via estrogen receptor β (ERβ). However, fulvestrant, an anti-estrogen-based endocrine therapeutic treatment, shows limited effects for non-small cell lung cancer (NSCLC) in phase II clinical trials. G protein-coupled estrogen receptor (GPER), a third estrogen receptor that binds to estrogen, has been found to be activated by fulvestrant, stimulating the progression of breast, endometrial, and ovarian cancers. We here demonstrated that cytoplasm-GPER (cGPER) (80.49 %) and nucleus-GPER (53.05 %) were detected by immunohistochemical analysis in NSCLC samples. cGPER expression was related to stages IIIA-IV, lymph node metastasis, and poorly differentiated NSCLC. Selective agonist G1 and 17β-estradiol (E2) promoted the GPER-mediated proliferation, invasion, and migration of NSCLC cells. Additionally, in vitro administration of E2 and G1 increased the number of tumor nodules, tumor grade, and tumor index in a urethane-induced adenocarcinoma model. Importantly, the pro-tumorigenic effects of GPER induced by E2 were significantly reduced by co-administering the GPER inhibitor G15 and the ERβ inhibitor fulvestrant, as compared to administering fulvestrant alone both in vitro and in vivo. Moreover, the phosphorylation of MAPK and Akt was involved in E2/G1-induced GPER activation. In conclusion, our results indicated that a pro-tumor function of GPER exists that mediated E2-/G1-dependent NSCLC progression and showed better efficiency regarding the co-targeting of GPER and ERβ, providing a rationale for further investigation of anti-estrogen clinical therapy.
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 10 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 20 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 50 mg (per vial)
Supelco
Urethane, analytical standard
Sigma-Aldrich
Urethane, ≥99.0% (GC)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥95% (HPLC)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥99%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, suitable for cell culture, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide lithium salt from Saccharomyces cerevisiae, ≥95%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, Grade AA-1
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, purified by column chromatography, ≥99%
Sigma-Aldrich
Urethane, ≥99%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥98%, BioUltra, from yeast
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Propionitrile, 99%
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Propionitrile, purum, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis