Merck
  • Abacopteris penangiana exerts testosterone-induced benign prostatic hyperplasia protective effect through regulating inflammatory responses, reducing oxidative stress and anti-proliferative.
Abacopteris penangiana exerts testosterone-induced benign prostatic hyperplasia protective effect through regulating inflammatory responses, reducing oxidative stress and anti-proliferative.
Journal of ethnopharmacology (2014-09-28)
Xian Yang, Liuliu Yuan, Chaomei Xiong, Chunping Yin, Jinlan Ruan
ABSTRACT
Abacopteris penangiana (Hook.) Ching (AP) is a member of parathelypteris glanduligera and used in folk medicine for the treatment of blood circulation and blood stasis, edema and inflammation as recorded in the ″Chinese Materia Medica″. The purpose of this study was to investigate the effects of total flavanol glycosides (TFA) from AP and its acid hydrolysate (AHT) on testosterone-induced benign prostatic hyperplasia (BPH) in rats by measuring the levels of inflammatory responses, oxidative stress and prostate cell proliferation. BPH was induced in rats by subcutaneous injection of testosterone after castration. Seventy rats were divided into seven groups. After oral administration of AHT and TFA (100 or 200mg/kg/d) for 4 weeks, the prostate index (PI), 5a-reductase (5α-R) and dihydrotestosterone (DHT) were determined. Then the activities of superoxide dismutase (SOD), glutathione peroxidase (GPx) and catalase (CAT) were determined. In addition, the relative inflammatory factors, cyclooxygenase-2 (COX-2), tumor necrosis factor-α (TNF-α), interleukin 1β (IL-1β), interleukin 6 (IL-6), interleukin 8 (IL-8) and interleukin 17 (IL-17) were measured. Finally, the prostatic expression of nuclear transcription factor-κB (NF-κB) and phosphoinositide3-kinase (PI3K)/Akt were determined by immunohistochemistry. The prostatic expression of Bcl-2 was determined by western blot analysis. The results showed that AHT and TFA decreased serum DHT and 5α-R activities compared with model group, as well as the PI and histopathological examination findings. In addition, oral treatment of AHT and TFA can significantly increase the activities of SOD, GPx and CAT while the level of MDA was significantly decreased compared with the model group. Moreover, AHT and TFA remarkably decreased the levels of inflammatory cytokines in prostatic tissue. Further investigation demonstrated that AHT and TFA treatment down-regulated the protein expressions of p-Akt, NF-κB and Bcl-2. These results suggest that AHT and TFA have anti-BPH properties via anti-inflammatory, antioxidant and anti-proliferative effects. Hence, AP represents a potential herb for the treatment of BPH.
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Finasteride, ≥98% (HPLC), powder
Finasteride, European Pharmacopoeia (EP) Reference Standard
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
4,4′-Diaminodiphenylmethane, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
4,4′-Diaminodiphenylmethane, ≥97.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Glutathione, European Pharmacopoeia (EP) Reference Standard
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Finasteride for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri