Merck
  • Association of histamine N-methyltransferase Thr105Ile polymorphism with Parkinson's disease and schizophrenia in Han Chinese: a case-control study.
Association of histamine N-methyltransferase Thr105Ile polymorphism with Parkinson's disease and schizophrenia in Han Chinese: a case-control study.
PloS one (2015-03-15)
Xinglong Yang, Chuanxin Liu, Jinxiang Zhang, Hongying Han, Xiuyan Wang, Zhoulin Liu, Yanming Xu
ABSTRACT
Parkinson's disease (PD) and schizophrenia (SCZ) are frequent central nervous disorders that have unclear etiologies but that show similarities in their pathogenesis. Since elevated histamine levels in the brain have been associated with PD and SCZ, we wanted to explore whether the Thr105Ile substitution in the histamine N-methyltransferase gene (HNMT-Thr105Ile), which impairs histamine degradation, is associated with either disease. We used the ligase detection reaction to genotype a case-control cohort of Han Chinese patients with PD or SCZ and healthy controls at the HNMT-Thr105Ile locus. The Ile allele was associated with reduced risk of PD (OR 0.516, 95% CI 0.318 to 0.838, p = 0.007) and of SCZ (OR 0.499, 95% CI 0.288 to 0.865, p = 0.011). Genotype frequencies and minor allele frequencies were similar between patients and controls when we compared males with females or early-onset patients with late-onset ones. Genotype and allele frequencies were not significantly different between PD patients with dyskinesia and PD patients without dyskinesia. Our results suggest that the heterozygous Thr/Ile genotype at the HNMT-Thr105Ile locus and the minor Ile105 allele protect against PD and SCZ in Han Chinese.
Sigma-Aldrich
Potassium acetate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium acetate, puriss., meets analytical specification of Ph. Eur., BP, E261, 99-101%
Sigma-Aldrich
Potassium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 10 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 20 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 50 mg (per vial)
Sigma-Aldrich
Potassium acetate, 99.98% trace metals basis
Sigma-Aldrich
Potassium acetate, 98%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥95% (HPLC)
Sigma-Aldrich
Potassium acetate, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥99%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, suitable for cell culture, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide lithium salt from Saccharomyces cerevisiae, ≥95%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, Grade AA-1
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, purified by column chromatography, ≥99%
Sigma-Aldrich
Potassium acetate, meets USP testing specifications
Sigma-Aldrich
Potassium acetate, Molecular Biology, ≥99.0%
Sigma-Aldrich
Potassium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥98%, BioUltra, from yeast
Sigma-Aldrich
Potassium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium acetate solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Potassium acetate solution, BioUltra, Molecular Biology, 5 M in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O