Merck
  • Glucuronidation of fimasartan, a new angiotensin receptor antagonist, is mainly mediated by UGT1A3.
Glucuronidation of fimasartan, a new angiotensin receptor antagonist, is mainly mediated by UGT1A3.
Xenobiotica; the fate of foreign compounds in biological systems (2014-07-19)
Eun-Sook Jeong, Yang-Weon Kim, Hyo-Ji Kim, Ho-Jung Shin, Jae-Gook Shin, Kwang Hee Kim, Yong Ha Chi, Soo Heui Paik, Dong-Hyun Kim
ABSTRACT
1. Fimasartan is an angiotensin receptor II antagonist used to treat patients with hypertension. This drug is mainly excreted into bile as either the parent compound or a glucuronide conjugate. In this study, we examined the glucuronidation of fimasartan and characterized the UDP-glucuronosyltransferases (UGTs) responsible for the glucuronidation. 2. Only one type of fimasartan glucuronide was observed after incubation with pooled human liver microsomes (HLMs) and was identified as an N2-glucuronide based on comparison with an authentic standard. 3. Among the 12 UGT isoforms tested, UGT1A1, UGT1A3 and UGT2B7 showed catalytic activity toward fimasartan glucuronidation. The intrinsic clearance (CLint) of UGT1A3 was 68.5- and 21.4-fold higher than that of UGT1A1 and UGT2B7, respectively, and the estimated relative contribution of UGT1A3 in human liver was 94.1%. Both chemical inhibition and correlation studies demonstrated that fimasartan glucuronidation activity in HLMs was significantly related with UGT1A3 activity. Fimasartan glucuronide was identified as a substrate for P-glycoprotein (Pgp) and breast cancer response protein (BCRP). 4. These findings collectively indicate that UGT1A3 is the major UGT isoform responsible for the glucuronidation of fimasartan, and this glucuronide is excreted from hepatocytes via MDR1 and BCRP.
Product Description
Supelco
Methanol, analytical standard
Sigma-Aldrich
Uridine, ≥99%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Uridine, BioUltra, ≥99%
Sigma-Aldrich
Uridine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Telmisartan for system suitability, European Pharmacopoeia (EP) Reference Standard
Telmisartan for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2,6-Diisopropylphenol, 97%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Bilirubin, purum, ≥95.0% (UV)
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
Uridine 5′-diphosphoglucuronic acid ammonium salt, 98-100%
Sigma-Aldrich
Niflumic acid