Merck
  • Use of compounded dispersing media for extemporaneous pediatric syrups with candesartan cilexetil and valsartan.
Use of compounded dispersing media for extemporaneous pediatric syrups with candesartan cilexetil and valsartan.
Acta pharmaceutica (Zagreb, Croatia) (2014-12-23)
Monika Musko, Malgorzata Sznitowska
ABSTRACT
Available tablets or capsules for adults are often used to prepare extemporaneously formulated medicines appropriate for children. The most acceptable drug forms in pediatric population are oral liquids and pharmacists use commercial dispersing media to compound syrups from an active substance or from tablets available on the market. In many countries ready-to-use dispersing media are not available or refunded, but pharmacists can use other compounded media, providing their compatibility and stability are proven. The aim of this study was to formulate and evaluate the stability of syrups with candesartan cilexetil (1 mg mL-1) and valsartan (4 mg mL-1) extemporaneously prepared using commercial tablets (Diovan® and Atacand®). The following three different suspending media, which could be easily made in a pharmacy, were investigated: V1 - with xanthan gum (0.5 %), V2 - the USP/NF vehicle for oral solution and V3 - the medium based on a simple sucrose syrup. The stability of preparations was studied during 35 days of storage in a dark place at controlled temperature of 25 and 4 °C. During the study, microscopic observation was carried out and pH, viscosity, and concentration of candesartan cilexetil and valsartan were analyzed. Syrups with valsartan prepared with V2 and V3 media were stable for 3 or 4 weeks when stored at 25 °C, while syrups with candesartan were stable for as long as 35 days. For syrups prepared using V1 medium, the 14-day expiry date was not achieved because of microbial deterioration.
Product Description
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Supelco
Methanol, analytical standard
Sigma-Aldrich
Candesartan cilexetil, ≥98% (HPLC)
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Candesartan cilexetil, European Pharmacopoeia (EP) Reference Standard
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis