Merck
  • A novel approach for high sensitive determination of sulfur mustard by derivatization and isotope-dilution LC-MS/MS analysis.
A novel approach for high sensitive determination of sulfur mustard by derivatization and isotope-dilution LC-MS/MS analysis.
Talanta (2014-12-06)
Bin Xu, Cheng Zong, Zhiyong Nie, Lei Guo, Jianwei Xie
ABSTRACT
A new isotope-dilution liquid chromatography-tandem mass spectrometry (LC-MS/MS) method for determination of sulfur mustard (SM) has been developed using a direct chemical derivatization method by nucleophile potassium thioacetate (PTA) in aqueous solution. The reaction conditions for derivatization, such as reaction temperature, time, solvent and concentration of PTA, were optimized for high performance. Reversed phase liquid chromatography was suitable for analysis of such a PTA derivatized SM in complex environmental samples. Compared with other conventional gas chromatography or gas chromatography-mass spectrometry methods for direct detection on SM, better sensitivity and selectivity were achieved by this direct derivatization and LC-MS/MS method, where SM can be detected as low as 0.05 ng/mL in acetonitrile. The linear range was from 0.1 to 1000 ng/mL. The relative standard deviation (RSD) of the intra-day precision was less than 11.8%, and RSD of the inter-day precision was less than 12.3%. The whole procedure for both derivatization and analysis was quick and simple, and the total time was less than 1h. This established method has been successfully employed for determination of spiking samples both in water and soil. A detection limit of 0.1 ng/mL was achieved for river water, while the SM in soil sample could be detected at 0.1 ng/g.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Potassium thioacetate, 98%
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione, 98%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), ≤12 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 35 μm particle size
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 200 μm particle size
Supelco
Hexane, analytical standard
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), 1 μm particle size
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O