Merck
  • Evaluation of ibuprofen loaded solid lipid nanoparticles and its combination regimens for pancreatic cancer chemoprevention.
Evaluation of ibuprofen loaded solid lipid nanoparticles and its combination regimens for pancreatic cancer chemoprevention.
International journal of oncology (2015-02-06)
Arvind Thakkar, Sushma Chenreddy, Jeffrey Wang, Sunil Prabhu
ABSTRACT
The objective of the present study was to establish the individual and combined chemopreventive potential of a widely used non-steroidal anti-inflammatory drug, ibuprofen (IBU), encapsulated in solid lipid nanoparticles (SLNs) for the chemoprevention of pancreatic cancer. The IBU SLNs were optimized using various lipids (Stearic acid, Compritol 888 ATO and Tripalmitin) and surfactants (Poloxamer 188, Tween-80). The synergistic effect of combination of IBU with sulforaphane (SFN) was also evaluated. Cell viability studies were conducted followed by colony formation and NF-κB DNA binding assays. The IC50 concentration of free IBU in human pancreatic cancer Panc-1 and MIA PaCa-2 cells were 1.25 and 1.26 mM, respectively. SLN optimization study of IBU revealed stearic acid (1:2 drug to lipid ratio) formulated with Poloxamer 188 to be the most efficacious in cell viability study. Upon encapsulation in SLNs, IC50 concentration of IBU-SLN was 113.8 and 122.6 µM for Panc-1 and MIA PACa-2 cells, respectively, reflecting a 10-fold reduction compared to free IBU. Combinations of low doses of free IBU (250 µM) and SFN (5 µM) reduced cell viability by ~55% (P<0.01), whereas a lower dose of encapsulated IBU-SLN (62.5 µM) and free SFN (5 µM) reduced cell viability by ~80% (P<0.001) for both Panc-1 and MIA PaCa-2 cells. These results reflect 4-fold reduction in IBU-SLN dose in combination compared to free IBU. Moreover, IBU-SLN and free SFN combination reduced number of colonies by ~50% (P<0.01). Further, IBU-SLN and SFN combinations showed down-regulation of DNA binding activity of the p50 subunit of NF-κB. In conclusion, these preliminary results demonstrate the potential of IBU as a chemopreventive agent against pancreatic cancer. Furthermore, when encapsulated in nanotechnology-based SLN delivery systems and delivered in combination with SFN provide evidence of a promising approach for pancreatic cancer prevention and therapy.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
5-(N-Methyl-N-isobutyl)­amiloride, ≥98% (TLC), powder
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Acetonitrile, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Phosphoric acid, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, 85.0-88.0%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG