Merck
  • A NMR-based, non-targeted multistep metabolic profiling revealed L-rhamnitol as a metabolite that characterised apples from different geographic origins.
A NMR-based, non-targeted multistep metabolic profiling revealed L-rhamnitol as a metabolite that characterised apples from different geographic origins.
Food chemistry (2014-12-23)
Satoru Tomita, Tadashi Nemoto, Yosuke Matsuo, Toshihiko Shoji, Fukuyo Tanaka, Hiroyuki Nakagawa, Hiroshi Ono, Jun Kikuchi, Mayumi Ohnishi-Kameyama, Yasuyo Sekiyama
ABSTRACT
This study utilises (1)H NMR-based metabolic profiling to characterise apples of five cultivars grown either in Japan (Fuji, Orin, and Jonagold) or New Zealand (Fuji, Jazz, and Envy). Principal component analysis (PCA) showed a clear separation between the Fuji-Orin-Jonagold class and the Jazz-Envy class, primarily corresponding to the differences in sugar signals, such as sucrose, glucose, and fructose. Multistep PCA removed the influence of dominant sugars and highlighted minor metabolites such as aspartic acid, 2-methylmalate, and an unidentified compound. These minor metabolites separated the apples into two classes according to different geographical areas. Subsequent partial least squares discriminant analysis (PLS-DA) indicated the importance of the unidentified metabolite. This metabolite was isolated using charcoal chromatography, and was identified as L-rhamnitol by 2D NMR and LC/MS analyses. The remarkable contribution of L-rhamnitol to geographic discrimination suggests that apples may be characterised according to various factors, including storage duration, cultivation method, and climate.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥97%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Deuterium oxide, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Deuterium oxide, 70 atom % D
Sigma-Aldrich
Deuterium oxide, 60 atom % D
Sigma-Aldrich
Acetonitrile, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitrile, analytical standard
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt