Merck
  • Enhanced invasion of lung adenocarcinoma cells after co-culture with THP-1-derived macrophages via the induction of EMT by IL-6.
Enhanced invasion of lung adenocarcinoma cells after co-culture with THP-1-derived macrophages via the induction of EMT by IL-6.
Immunology letters (2014-04-05)
Che Dehai, Pan Bo, Tian Qiang, Shang Lihua, Liu Fang, Jin Shi, Cao Jingyan, Yu Yan, Wang Guangbin, Yuan Zhenjun
ABSTRACT
Lung cancer is the leading cause of cancer mortality worldwide, and the cause of death is metastasis. The epithelial-to-mesenchymal transition (EMT) plays a key role in the process of metastasis. Macrophages within the lung cancer microenvironment release cytokines, such as interleukin-6 (IL-6), and promote lung cancer cell invasion and metastasis. However, the interaction between macrophages and lung cancer cells and the effect of this interaction on the expression of IL-6, EMT, and the invasiveness of lung cancer cells remain unclear. Therefore, we established an in vitro co-culture model of human lung adenocarcinoma A549 or H1299 cells with THP-1-derived macrophages to illuminate the important role of macrophages in the invasion of lung cancer. In this study, we demonstrated that the concentrations of IL-6 in the co-culture supernatants were significantly increased compared with controls. Thus, a complex chemical cross-talk is induced by the indirect cell-to-cell contact between lung cancer cells and THP-1-derived macrophages. THP-1-derived macrophages appeared to play an important initiator role in the process. The analysis of the mRNA expression profiles of the sorted cells from the co-culture system revealed that the co-cultured lung cancer cells are the main source of the observed increase in IL-6 secretion. In addition, the interactions between lung cancer cells and THP-1-derived macrophages are bidirectional. The THP-1-derived macrophages underwent differentiation towards the M2-macrophage phenotype during the co-culture process. The expression of IL-6 was correlated with the induction of EMT, which contributed to a significant increase in the invasiveness of the A549 and H1299 cells in vitro. In addition, the addition of an anti-IL-6 antibody reversed these changes. In summary, we demonstrated that the in vitro co-culture of A549 or H1299 cells with THP-1-derived macrophages upregulates IL-6 expression, which increases the invasion ability of the A549 and H1299 cells through the EMT pathway. The THP-1-derived macrophages that interacted with the lung cancer cells differentiated towards the M2-macrophage phenotype. Thus, the inhibition of IL-6 or of the interactions between lung cancer cells and macrophages may be an effective target for anti-cancer therapy in patients with non-small cell lung cancer.
Sigma-Aldrich
MISSION® esiRNA, targeting human VIM
Sigma-Aldrich
MISSION® esiRNA, targeting human CDH1
Sigma-Aldrich
MISSION® esiRNA, targeting human IL6
Sigma-Aldrich
E-Cadherin human, recombinant, expressed in E. coli, 0.5 mg protein/mL
Sigma-Aldrich
MISSION® esiRNA, targeting human CD68, RP11-186B7.4
Sigma-Aldrich
Interleukin-6 human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture