Merck
  • New flavonoids from Campylotropis hirtella with immunosuppressive activity.
New flavonoids from Campylotropis hirtella with immunosuppressive activity.
Fitoterapia (2014-04-09)
Xiaoping Li, Bixia Xuan, Qingyao Shou, Zhengwu Shen
ABSTRACT
In an effort to identify natural compounds with immunosuppressive activity, nine new flavonoids, including one isoflav-3-ene derivative (1), one coumaronochromone (2), two isoflavanones (3, 4), one isoflavone derivative (6), one isoflavone (7), three flavonols (8, 9, 10), as well as one known compound, hydroisoflavone C (5), were isolated from the roots of Campylotropis hirtella. The structures of these compounds were elucidated by extensive spectroscopic measurements. All of the compounds were assessed for immunosuppressive activity. Among the isolates, compound 2 showed good inhibitory activity against mitogen-induced splenocyte proliferation with an IC50 of 0.28 μM and relatively low cytotoxicity.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ≥99.9% (GC)
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ultrapure Acetonitrile
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material