Merck
  • Passive and iontophoretic transport of fluorides across enamel in vitro.
Passive and iontophoretic transport of fluorides across enamel in vitro.
Journal of pharmaceutical sciences (2014-04-10)
Wei Ren, Arif Baig, S Kevin Li
ABSTRACT
Passive and iontophoretic transport of fluoride from three fluoride sources, NaF, sodium monofluorophosphate (MFP), and SnF2 solutions, across bovine enamel was investigated to (1) determine the characteristics of the intrinsic barrier of enamel for ion transport, (2) examine the feasibility of iontophoretically enhanced transport of fluoride across enamel, and (3) identify the transport mechanisms involved in enamel iontophoresis. Conductivity experiments were performed with bovine enamel specimens in side-by-side diffusion cells to evaluate the electrical and barrier properties of the enamel with electrolytes of different ion sizes and under different ion concentrations and pH conditions in vitro. Transport experiments of the enamel were performed in the diffusion cells with the NaF, MFP, and SnF2 solutions. The conductivity results showed that the enamel specimens behaved as a neutral membrane or that of low pore charge density. Cathodal iontophoresis significantly enhanced the delivery of fluoride ions across the enamel from the solutions over passive transport, consistent with Nernst-Planck theory and the direct field effect (i.e., electrophoresis) as the dominant flux-enhancing mechanism. The enamel demonstrated significant transport hindrance for the ions, and the effective pore radii of the transport pathways in the enamel were found to be approximately 0.7-0.9 nm.
Sigma-Aldrich
Potassium chloride, Molecular Biology, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Potassium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Supelco
Potassium chloride solution, ~3 M KCl, saturated with silver chloride
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Potassium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture