Merck
  • Neurons have an active glycogen metabolism that contributes to tolerance to hypoxia.
Neurons have an active glycogen metabolism that contributes to tolerance to hypoxia.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism (2014-02-27)
Isabel Saez, Jordi Duran, Christopher Sinadinos, Antoni Beltran, Oscar Yanes, María F Tevy, Carlos Martínez-Pons, Marco Milán, Joan J Guinovart
ABSTRACT
Glycogen is present in the brain, where it has been found mainly in glial cells but not in neurons. Therefore, all physiologic roles of brain glycogen have been attributed exclusively to astrocytic glycogen. Working with primary cultured neurons, as well as with genetically modified mice and flies, here we report that-against general belief-neurons contain a low but measurable amount of glycogen. Moreover, we also show that these cells express the brain isoform of glycogen phosphorylase, allowing glycogen to be fully metabolized. Most importantly, we show an active neuronal glycogen metabolism that protects cultured neurons from hypoxia-induced death and flies from hypoxia-induced stupor. Our findings change the current view of the role of glycogen in the brain and reveal that endogenous neuronal glycogen metabolism participates in the neuronal tolerance to hypoxic stress.
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Uridine, BioUltra, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Uridine, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Uridine, powder, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Pricing and availability is not currently available.
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Pricing and availability is not currently available.