Merck
  • Secretory clusterin is upregulated in rats with pulmonary arterial hypertension induced by systemic-to-pulmonary shunts and exerts important roles in pulmonary artery smooth muscle cells.
Secretory clusterin is upregulated in rats with pulmonary arterial hypertension induced by systemic-to-pulmonary shunts and exerts important roles in pulmonary artery smooth muscle cells.
Acta physiologica (Oxford, England) (2014-07-30)
X Liu, L Meng, J Li, J Meng, X Teng, H Gu, S Hu, Y Wei
ABSTRACT
Phenotype modification of pulmonary artery smooth muscle cells (PASMCs) (excessive proliferation, migration and impaired apoptosis) plays central roles in pulmonary vascular remodelling of pulmonary arterial hypertension (PAH); however, the potential mechanism and contributing factors involved in the phenotype alteration in PASMCs are still not completely elucidated. This study attempted to investigate the expression pattern of secretory clusterin (sCLU), a prosurvival protein, in systemic-to-pulmonary shunt-induced PAH rats and the potential roles of sCLU in pulmonary vascular remodelling. An original rat model of systemic-to-pulmonary shunt-induced PAH was established by combined surgery as we previously reported. Lung tissues were harvested at specific time points for real-time polymerase chain reaction, Western blot and immunohistochemisty analysis; meanwhile, plasma was collected for enzyme-linked immunosorbent assay. Cell culture experiments were performed using cultured human PASMCs (HPASMCs). Expression of sCLU was significantly increased in lungs exposed to systemic-to-pulmonary shunt. Moreover, plasma sCLU levels were markedly elevated with the progression of PAH in rats and also presented a positive correlation with pulmonary hemodynamic indices. In vitro cell culture assay indicated that sCLU expression and secretion increased with the phenotype modification of HPASMCs; furthermore, sCLU promoted HPASMCs proliferation, migration and apoptosis resistance, at least in part, via Erk1/2 and Akt signalling pathways. These results demonstrate that sCLU is functionally an important phenotype modulator of PASMCs, and its upregulation in lung tissues may exert a deteriorative role in pulmonary vascular remodelling.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, absolute, sales not in Germany, ≥99.8% (vol.)
Sigma-Aldrich
Propidium iodide solution
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology