Merck
  • Hypoxia-inducible factors modulate the stemness and malignancy of colon cancer cells by playing opposite roles in canonical Wnt signaling.
Hypoxia-inducible factors modulate the stemness and malignancy of colon cancer cells by playing opposite roles in canonical Wnt signaling.
PloS one (2014-11-15)
Paula Santoyo-Ramos, María Likhatcheva, Eduardo A García-Zepeda, M Cristina Castañeda-Patlán, Martha Robles-Flores
ABSTRACT
This study examined the role played by hypoxia-inducible factors (HIFs) in malignant phenotype maintenance and canonical Wnt signaling. Under normoxia, we determined that both HIF-1α and HIF-2α are expressed in human colon cancer cells but not in their non-malignant counterparts. The stable knockdown of HIF-1α or HIF-2α expression induced negative effects on the malignant phenotype of colon cancer cells, with lactate production, the rate of apoptosis, migration, CXCR4-mediated chemotaxis, and tumorigenic activity all being significantly affected by HIF knockdown and with HIF-1α depletion exerting greater effects. Knockdown of these two HIF transcripts induced different and even opposite effects on β-catenin transcriptional activity in colon cancer cells with different genetic Wnt signaling pathways. In SW480 cells, HIF-2α knockdown did not affect β-catenin levels, increasing the transcriptional activity of β-catenin by inducing its nuclear accumulation, whereas HIF-1α silencing negatively affected the stability and transcriptional activity of β-catenin, inducing its exit from the nuclei and its recruitment to the cell membrane by E-cadherin. In addition, although HIF-1α depletion induced a reversal of the epithelial-to-mesenchymal transition (EMT), HIF-2α silencing altered the expression of the stem cell markers CD44, Oct4, and CD24 and of the differentiation marker CK20 in the opposite direction as HIF-1α silencing. Remarkably, HIF-2α knockdown also enhanced β-catenin transcriptional activity under hypoxia in cells that displayed normal Wnt signaling, suggesting that the gene negatively modulates canonical Wnt signaling in colon cancer cells. Taken together, our results indicate that HIFs play opposing roles in canonical Wnt signaling and are essential for the stemness and malignancy maintenance of colon cancer cells.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material